Skip to Content
Merck
All Photos(1)

Key Documents

MAB1675

Sigma-Aldrich

Anti-Cytokeratin 19 Antibody, clone E6

ascites fluid, clone E6, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

E6, monoclonal

species reactivity

fish, rat, rabbit

should not react with

human, mouse

packaging

antibody small pack of 25 μL

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT19(3880)
mouse ... Krt19(16669)
rat ... Krt19(360626)

Specificity

Rat keratin 19 (40kD), analogous to human keratin 19.

Immunogen

Keratin from rat colon.

Application

Anti-Cytokeratin 19 Antibody, clone E6 detects level of Cytokeratin 19 & has been published & validated for use in WB, IH.
Immunohistochemistry frozen tissue sections.

Immunoblotting

Optimal working dilutions must be determined by end user.

Quality

Tested

Other Notes

Please refer to lot specific datasheet.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Houda Darwiche et al.
Hepatic medicine : evidence and research, 3, 89-98 (2011-09-20)
BACKGROUND AND AIMS: Activation of the oval cell compartment occurs in the liver when hepatocytes are functionally compromised and/or unable to divide. Our goal was to investigate the systemic signals responsible for determining the efficiency of oval cell-mediated liver regeneration
P J Salas et al.
The Journal of cell biology, 137(2), 359-375 (1997-04-21)
In a previous publication (Rodriguez, M.L., M. Brignoni, and P.J.I. Salas. 1994. J. Cell Sci. 107: 3145-3151), we described the existence of a terminal web-like structure in nonbrush border cells, which comprises a specifically apical cytokeratin, presumably cytokeratin 19. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service