Skip to Content
Merck
All Photos(2)

Documents

MAB1567A4

Sigma-Aldrich

Anti-MAG Antibody, clone 513, Alexa Fluor 488 Conjugate

clone 513, from mouse, ALEXA FLUOR 488

Synonym(s):

Myelin-associated glycoprotein, Siglec-4a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

513, monoclonal

species reactivity

rat, mouse

species reactivity (predicted by homology)

chicken (based on 100% sequence homology), human (based on 100% sequence homology), frog (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MAG(4099)

General description

Myelin-associated glycoprotein (MAG), a member of the immunoglobulin superfamily, is a membrane glycoprotein of 100 kDa, and is thought to be involved in the process of myelination. Axonal regeneration in the adult central nervous system (CNS) is limited by two proteins in myelin, Nogo and MAG. MAG has been implicated in the inhibition of nerve regeneration in the CNS.

Immunogen

L2 epitope carrying glycoprotein extract of 1-2 day old chicken brain

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected MAG in rat E18 primary cortex cells.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
This Anti-MAG Antibody, clone 513, Alexa Fluor 488 Conjugate is validated for use in IC, IH for the detection of MAG.

Quality

Evaluated by Immunohistochemistry in adult mouse brain tissue.

Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected MAG in adult mouse brain tissue.

Target description

69 kDa calculated

Physical form

Protein A purified
Purified mouse monoclonal IgG1 conjugated to Alexa Fluor 488 in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Adult mouse brain tissue

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yevgeniya A Mironova et al.
eLife, 5 (2016-03-24)
Proper development of the CNS axon-glia unit requires bi-directional communication between axons and oligodendrocytes (OLs). We show that the signaling lipid phosphatidylinositol-3,5-bisphosphate [PI(3,5)P2] is required in neurons and in OLs for normal CNS myelination. In mice, mutations of Fig4, Pikfyve
Allison Y Louie et al.
Journal of neuroinflammation, 20(1), 190-190 (2023-08-19)
Recent data suggest that myelin may be altered by physiological events occurring outside of the central nervous system, which may cause changes to cognition and behavior. Similarly, peripheral infection by non-neurotropic viruses is also known to evoke changes to cognition
Lizhao Feng et al.
iScience, 26(11), 108037-108037 (2023-10-23)
Myelin defects cause a collection of myelin disorders in the brain. The lack of human models has limited us from better understanding pathological mechanisms of myelin diseases. While human induced pluripotent stem cell (hiPSC)-derived spheroids or organoids have been used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service