Skip to Content
Merck
All Photos(1)

Key Documents

MAB1160

Sigma-Aldrich

Anti-gC1q-R/p33 Antibody, CT, aa 204-218, clone 74.5.2

clone 74.5.2, Chemicon®, from mouse

Synonym(s):

ASF/SF2-associated protein p32, Glycoprotein gC1qBP, Hyaluronan-binding protein 1, Mitochondrial matrix protein p32, gC1q-R protein, p33, C1qBP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

74.5.2, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... C1QBP(708)

Specificity

Recognizes the mature form and truncated forms of gC1q-R/p33 complement receptor. Antibody is directed against the carboxyl-terminus of the receptor (residues 204-218), a portion of the molecule specifies interaction with HK and Factor XII and blocks binding of HK to gC1q-R.

Immunogen

Epitope: C-terminus, aa 204-218
Recombinant human gC1q-R.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-gC1q-R/p33 Antibody, C-terminus, aa 204-218, clone 74.5.2 is validated for use in ELISA, FC, IP, WB for the detection of gC1q-R/p33.
Western blot

Immunoprecipitation

ELISA

FACS

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02M PBS pH 7.6, 0.25M NaCl with 0.1% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service