Skip to Content
Merck
All Photos(1)

Key Documents

LX0260

Supelco

Ligroine

Practical

Synonym(s):

Ligroin, Petroleum Ether, Petroleum Benzin, Mineral spirits, Petroleum spirits

Sign Into View Organizational & Contract Pricing

Select a Size

4 L
CZK 5,510.00

CZK 5,510.00


Estimated to ship onMay 26, 2025Details

This item is subject to Excise Duty. Please check the cart for more details.

Request a Bulk Order

Select a Size

Change View
4 L
CZK 5,510.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12191502
EC Index Number:
232-453-7
bp:
60-80 °C (lit.)

CZK 5,510.00


Estimated to ship onMay 26, 2025Details

This item is subject to Excise Duty. Please check the cart for more details.

Request a Bulk Order

vapor density

>1 (vs air)

Quality Level

form

liquid

color

APHA: ≤10

refractive index

n20/D 1.376 (lit.)

bp

60-80 °C (lit.)

transition temp

flash point -7 °C

density

0.656 g/mL at 25 °C (lit.)
0.67-0.69  g/mL at 25 °C

shipped in

ambient

storage temp.

room temp

SMILES string

[Mg+]c1ccc(cc1)C.[Br-]

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Practical

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Boiling range60-90°CColor (APHA)10 maxDensity at 25°C0.67-0.69g/mLFormClear liquidInfrared SpectrumConforms to standard

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service