Skip to Content
Merck

KWLCA02FF3

Opticap® XL 2 Capsule Media: Milligard® LPB 1.2 / 0.5 µm Nominal

Chemistry: Low Protein-Binding Mixed Cellulose Esters (MCE) Inlet: 3/4 in. Sanitary Flange Outlet: 3/4 in. Sanitary Flange Max Operating Pressure: 80 psi @ 25 °C

Synonym(s):

Opticap XL2 Milligard Low Protein Binding 1.2/0.5 μm 3/4 in. TC/TC, Milligard® LPB 1.2/0.5 µm nominal, Opticap® XL Capsule

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32031610
UNSPSC Code:
23151806
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Opticap® XL 2 Capsule Media: Milligard® LPB 1.2 / 0.5 µm Nominal, Chemistry: Low Protein-Binding Mixed Cellulose Esters (MCE) Inlet: 3/4 in. Sanitary Flange Outlet: 3/4 in. Sanitary Flange Max Operating Pressure: 80 psi @ 25 °C

Quality Segment

material

mixed cellulose esters (MCE) (low protein-binding), polyester support, polypropylene , polypropylene housing, polypropylene vent cap, silicone seal

reg. compliance

meets FDA Indirect Food Additive requirements cited in 21 CFR 177-182 (all component materials)

sterility

non-sterile

sterilization compatibility

autoclavable compatible

product line

Opticap® XL 2

feature

hydrophilic

manufacturer/tradename

Opticap®

parameter

1.0 bar max. inlet pressure (15 psi) at 80 °C, 2.75 bar max. inlet pressure (40 psi) at 60 °C, 25 °C max. inlet temp., 3.5 bar max. differential pressure (50 psid) at 25 °C, 5.5 bar max. inlet pressure (80 psi) at 23 °C, 80 psig max. inlet pressure

technique(s)

prefiltration: suitable

L

14.2 cm (5.6 in.)

W

3.3 in.

cartridge nominal length

2 in. (5 cm)

diam.

12.5 cm (4.9 in.)

filtration area

0.07 m2

inlet connection diam.

3/4 in.

inlet to outlet W

14.2 cm (5.6 in.)

outlet connection diam.

3/4 in.

impurities

≤0.5 EU/mL bacterial endotoxins (LAL test, Aqueous extraction), <0.5 EU/mL USP bacterial endotoxins (LAL test, sample aqueous extraction)

gravimetric extractables

≤60 mg/capsule

matrix

Milligard® LPB

pore size

1.2/0.5 μm nominal pore size, 1.2/0.5 μm pore size

input

sample type liquid

fitting

inlet sanitary flange, outlet sanitary flange, (19 mm (3/4 in.) Sanitary Flange Inlet and Outlet)

General description

Device Configuration: Capsule

Features and Benefits

Filter Grade: Low Protein Binding Milligard® Media
Format: Double Layer

Packaging

Double Easy-Open bag

Preparation Note

Sterilization Method
3 autoclave cycles of 30 min @ 121 °C; not in-line steam sterilizable
This product was manufactured with a media combination which meets the criteria for a "non-fiber releasing" filter as defined in 21 CFR 210.3 (b) (6).

Analysis Note

Gravimetric Extractables: after a 1 L flush and 24 hours in water at controlled room temperature
Will meet the USP Oxidizable Substances Test requirements after a water flush of ≥1 L

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: BioProcessing
  • Intended Use: Reduction or removal of microorganism/bioburden
  • Instructions for Use: Please refer installation section of OPTICAP® XL Capsules and OPTICAP® XLT Capsules user guide
  • Storage Statement: Store in dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Replaces: KWSCL4FF3
These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.

Legal Information

MILLIGARD is a registered trademark of Merck KGaA, Darmstadt, Germany
OPTICAP is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

EMD Millipore Corporation certifies that this product complies with the European Pressure Equipment Directive, 97/23/EC of 29 May 1997. This product has been classified under Article 3 § 3 of the Pressure Vessel Directive. It has been designed and manufactured in accordance with sound engineering practice to ensure safe use. In compliance with Article 3 § 3 of this Pressure Vessel Directive, this product does not bear the CE mark.


Still not finding the right product?


Storage Class

4.1B - Flammable solid hazardous materials

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library