HTS110M
ChemiSCREEN Membrane Preparation Recombinant Human 5-HT4B Serotonin Receptor
Human 5-HT4 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
General description
5-Hydroxytryptamine (5-HT, also commonly known as serotonin) is synthesized in enterochromaffin cells in the intestine and in serotonergic nerve terminals. In the periphery, 5-HT mediates gastrointestinal motility, platelet aggregation, and contraction of blood vessels. Many functions of the central nervous system are influenced by 5-HT, including sleep, motor activity, sensory perception, arousal and appetite. A family of 12 GPCRs and one ion channel mediate the biological effects of 5-HT (Hoyer et al., 1994). 5-HT4 comprises at least 8 isoforms varying at the C-terminus, which are generated by alternative splicing. The expression and distribution of these splice variants differs among organs and tissues with many of them present in several tissues such as atrium, brain, and GI tract (Bockaert et al. 2004). To date, all isoforms have been shown to activate adenylyl cyclase in vitro, and no difference in signal transduction between C-terminal 5-HT4 receptor variants has been demonstrated. 5-HT4 receptors are of potential interest for the treatment of patients with GI motility disorders and Alzheimer′s disease (De Maeyer et al. 2008; Lezoualc’h 2007). Millipore′s 5-HT4B membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of agonists and antagonists of 5-HT4B. The membrane preparations exhibit a Kd of 0.46 nM for [3H]-GR113808. With 1 nM [3H]-GR113808, 5 μg/well 5-HT4B Membrane Prep yields greater than 10 fold signal-to-background ratio.
Human HTR4, transcript b, encoding 5-HT4B
Application
Human 5-HT4 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Radioligand binding assay and GTPγS binding.
Biochem/physiol Actions
GPCR Class: A
Protein Target: 5-HT4
Target Sub-Family: Serotonin
Quality
Table 1. Signal:background and specific binding values obtained in a competition binding assay with varying amounts of 5-HT4B membrane prep.
SPECIFICATIONS: 1 unit = 5 μg membrane preparation
Bmax: 12.5 pmol/mg
Kd: 2.7 nM for [3H]-GR113808
10 µg/well | 5 µg/well | |
---|---|---|
Signal:Background | 26.4 | 21.7 |
Specific Binding (cpm) | 5286 | 4879 |
SPECIFICATIONS: 1 unit = 5 μg membrane preparation
Bmax: 12.5 pmol/mg
Kd: 2.7 nM for [3H]-GR113808
Specifications
Inucbation Conditions
RECOMMENDED ASSAY CONDITIONS: Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in assay buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with assay buffer. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with assay buffer. The plate is dried and counted.
Assay buffer: 25 mM Tris, pH 7.4, filtered and stored at 4°C
Radioligand: [3H]-GR113808 (Amersham TRK944)
One package contains enough membranes for at least 200 assays (units), where one unit is the amount of membrane that will yield greater than 10-fold signal:background with 3H-labeled GR113808 at 1 nM.
RECOMMENDED ASSAY CONDITIONS: Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in assay buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with assay buffer. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with assay buffer. The plate is dried and counted.
Assay buffer: 25 mM Tris, pH 7.4, filtered and stored at 4°C
Radioligand: [3H]-GR113808 (Amersham TRK944)
One package contains enough membranes for at least 200 assays (units), where one unit is the amount of membrane that will yield greater than 10-fold signal:background with 3H-labeled GR113808 at 1 nM.
Physical form
Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membrane protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.
Packaging method: Membrane protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.
Storage and Stability
Store at –70°C. Product is stable for at least 6 months from the date of receipt when stored as directed. Do not freeze and thaw.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of medicinal chemistry, 58(7), 3172-3187 (2015-03-21)
In this work, we describe the synthesis and in vitro evaluation of a novel series of multitarget-directed ligands (MTDL) displaying both nanomolar dual-binding site (DBS) acetylcholinesterase inhibitory effects and partial 5-HT4R agonist activity, among which donecopride was selected for further
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service