Skip to Content
Merck
All Photos(1)

Documents

HTS076RTA

Sigma-Aldrich

Ready-to-Assay PAF Platelet Activating Factor Receptor Frozen Cells

Human

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.84

product name

Ready-to-Assay PAF Platelet Activating Factor Receptor Frozen Cells, Human PAF GPCR frozen cells for Calcium Flux FLIPR Assays.

biological source

human

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

shipped in

dry ice

General description

Millipore’s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

Platelet-activating factor (PAF) is a bioactive phospholipid that was originally characterized by its ability to induce platelet aggregation, but was subsequently shown to mediate angiogenesis, inflammation and neural development. The PAF receptor is a G protein coupled receptor that signals through multiple pathways and mediates several cellular responses including cell motility, smooth muscle contraction, and cytokine and leukotriene release (Stafforini et al., 2003). Millipore’s cloned human PAF-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant PAF expression on the cell surface and contains high levels of the promiscuous G protein G15 to enhance coupling of the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for antagonists of interactions between PAF and its ligands.

Cell Line Description

Host cells: Chem-1

Biochem/physiol Actions

Protein Target: PAF
Target Sub-Family: Platelet Activating Factor

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service