Skip to Content
Merck

Skip To

HATF08550

Immobilon®-NC Membrane

85 mm disc, 0.45 µm pore size, Triton-free, mixed cellulose esters

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 pkg
Please contact Customer Service for Availability
5 730,00 CZK

About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602
NACRES:
NB.22
Material:
mixed cellulose esters (MCE) membrane (surfactant-free), nitrocellulose membrane, plain filter, white filter
Detection method:
chemiluminescent, colorimetric, fluorometric, radioactive
Pore size:
0.45 μm pore size
Compatibility:
for use with Amido black, for use with CPTS, for use with Colloidal gold, for use with India ink, for use with Ponceau-S red

5 730,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Immobilon®-NC Membrane, Triton-free, mixed cellulose esters, 0.45 m, 85 mm, disc, 0.45 um ( 0.45 micron filter ) pore size Mixed Cellulose Esters, Surfactant-Free, Transfer Membrane for western blotting using Chromogenic, Radioactive or Fluorescent detection methods.

Quality Segment

material

mixed cellulose esters (MCE) membrane (surfactant-free), nitrocellulose membrane, plain filter, white filter

feature

hydrophilic

manufacturer/tradename

Immobilon®

technique(s)

western blot: suitable

filter diam.

85 mm

thickness

180 μm

refractive index

n/D 1.51

pore size

0.45 μm pore size

capacity

117 μg/cm2 (insulin), 160 μg/cm2 adsorption capacity (BSA), 259 μg/cm2 adsorption capacity (goat IgG)

compatibility

for use with Amido black, for use with CPTS, for use with Colloidal gold, for use with India ink, for use with Ponceau-S red

detection method

chemiluminescent, colorimetric, fluorometric, radioactive

shipped in

ambient

General description

Filter Code: HATF
Primary Binding Mechanisms: Electrostatic, hydrophobic

Application

  • Biological solutions
  • Cell contact
  • Very small volumes requiring surfactant-free surfaces
  • Isolation of virus-like particles in wastewater
  • Microplastics analysis grade water
  • Nucleic acid binding, including eDNA

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HATF08250HATF02500HATF00010
description

0.45 um ( 0.45 micron filter ) pore size Mixed Cellulose Esters, Surfactant-Free, Transfer Membrane for western blotting using Chromogenic, Radioactive or Fluorescent detection methods.

description

0.45 um ( 0.45 micron filter ) pore size Mixed Cellulose Esters, Surfactant-Free, Transfer Membrane for western blotting using Chromogenic, Radioactive or Fluorescent detection methods.

description

25 mm diameter disc, 0.45 µm pore size, Triton-free, mixed cellulose esters transfer membrane

description

1 roll, 33 cm x 3 m, 0.45 µm pore size, Triton-free, mixed cellulose esters transfer membrane

pore size

0.45 μm pore size

pore size

0.45 μm pore size

pore size

0.45 μm pore size, 79 % porosity

pore size

0.45 μm pore size

Quality Level

400

Quality Level

400

Quality Level

400

Quality Level

400

detection method

colorimetric, chemiluminescent, radioactive, fluorometric

detection method

radioactive, chemiluminescent, colorimetric, fluorometric

detection method

chemiluminescent, fluorometric, colorimetric, radioactive

detection method

radioactive, colorimetric, chemiluminescent, fluorometric

material

nitrocellulose membrane, white filter, mixed cellulose esters (MCE) membrane (surfactant-free), plain filter

material

plain filter, mixed cellulose esters (MCE) membrane (surfactant-free), nitrocellulose membrane, white filter

material

nitrocellulose membrane, white filter, mixed cellulose esters (MCE) membrane (surfactant-free), plain filter

material

white filter, plain filter, nitrocellulose membrane, mixed cellulose esters (MCE) membrane

compatibility

for use with India ink, for use with Colloidal gold, for use with Ponceau-S red, for use with CPTS, for use with Amido black

compatibility

for use with Ponceau-S red, for use with CPTS, for use with Amido black, for use with India ink, for use with Colloidal gold

compatibility

for use with CPTS, for use with Amido black, for use with Colloidal gold, for use with India ink, for use with Ponceau-S red

compatibility

for use with Ponceau-S red, for use with Colloidal gold, for use with India ink, for use with Amido black, for use with CPTS

manufacturer/tradename

Immobilon®

manufacturer/tradename

Immobilon®

manufacturer/tradename

Immobilon®

manufacturer/tradename

Immobilon®


Still not finding the right product?


Pictograms

Flame

Signal Word

Danger

Hazard Codes

Hazard Classifications

Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters