Skip to Content
Merck

Skip To

GF330

IL-1α,
Human Recombinant Animal Free

Synonym(s):

IL1 alpha

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 μg

Estimated to ship onJune 08, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 10,200.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
eCl@ss:
32160405

CZK 10,200.00


Estimated to ship onJune 08, 2026Details


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

Quality Segment

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

General description

IL-1α is a non-secreted proinflammatory cytokine produced in a variety of cells including monocytes, tissue macrophages, keratinocytes and other epithelial cells. Both IL-1α and IL-1β binds to the same receptor and has similar if not identical biological properties. These cytokines have a broad range of activities including, stimulation of thymocyte proliferation, by inducing IL-2 release, B-cell maturation and proliferation, mitogenic FGF-like activity and the ability to stimulate the release of prostaglandin and collagenase from synovial cells. However, whereas IL-1β is a secreted cytokine, IL-1α is predominantly a cell-associated cytokine.
Recombinant human IL-1α is an 18.0 kDa protein containing 159 amino acid residues.
Product Source: Protein is expressed in E.coli.
The protein is manufactured without the use of any animal products/reagents and is considered Animal-Free and can be used in protocols where possible transmisson of animal viruses and contamination must be avoided.

Application

Research Category
Stem Cell Research
Research Sub Category
Growth Factors & Receptors

Physical form

Product is presented in 3.2 mM Tris, pH 8.3 + 16 mM NaCl and is filtered through a 0.2 micron filter before lyophilization.

Preparation Note

Store at -20°C for up to 4 months from date of receipt Centrifuge the vial prior to opening.
Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein and store in working aliquots at -20°C.

Analysis Note

Determined its ability to stimulate the proliferation of mouse D10S cells. The expected ED50 is ≤0.001 ng/ml, corresponding to a specific activity of ≥1 x10^9 units/mg.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF331GF324GF327
shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

UniProt accession no.

P01583

UniProt accession no.

P01584

UniProt accession no.

P18510

UniProt accession no.

P40933

impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

storage temp.

-

storage temp.

-10 to -25°C

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Storage Class

11 - Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters