FR180380623
Estapor® Fluorescent Carboxylated microspheres
F1-EU-010 FluoEU COOH PS 1%, Red Fluorescent carboxylated microspheres 160nm, Lateral Flow Immunoassay - For ordering, click "Request more information"
Synonym(s):
Detector Particles, Fluorescent Beads
Select a Size
CZK 14,500.00
Estimated to ship onFebruary 25, 2025Details
Select a Size
About This Item
CZK 14,500.00
Estimated to ship onFebruary 25, 2025Details
Recommended Products
Quality Level
packaging
pkg of 10-1000 mL (dark bottle)
manufacturer/tradename
Estapor®
concentration
1% solids
technique(s)
lateral flow assay: suitable (Immunoassay)
color
fluorescent red
matrix
polymeric Poly-styrene-co-acrylate
bead diameter
0.130-0.190 μm±<5%
capacity
>300 μeq/g surface charge density
λmax
350 nm (Emission at 613 nm)
functional group
carboxylic acid (COOH)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Estapor® Microspheres, for polymeric supports in diagnostics, life sciences, biotechnology, & environmental applications, offers 100+ microsphere types.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service