Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB208P

Sigma-Aldrich

Milli-Mark® Anti-CD79αcy-PE Antibody, clone HM57

clone HM57, Milli-Mark®, from mouse

Synonym(s):

B-cell antigen receptor complex-associated protein alpha chain, Ig-alpha, MB-1 membrane glycoprotein, Surface IgM-associated protein, Membrane-bound immunoglobulin-associated protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HM57, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD79A(973)

General description

CD79 is a disulphide-linked transmembrane heterodimer with a molecular mass of 82-95 kDa belonging to the immunoglobulin superfamily. It comprises the two glycoproteins, CD79α, with a molecular mass of 40-45 kDa, and CD79β, with a molecular mass of 37 kDa. CD79 is non-covalently associated with surface Ig, forming the B-cell receptor complex, which is required for antigen recognition. The expression of CD79 is largely restricted to B-lineage cells (Pilozzi,1998). In precursor B cells, the CD79 protein chains are already expressed in the cytoplasm (CyCD79). Surface expression of CD79 begins at the pro-B cell stage and persists throughout the B-cell differentiation. CD79α is more strongly expressed by B cells in the follicular mantle zone than by germinal centre B cells, suggesting that activation of mature B cells downregulates CD79 expression. CD79 expression ceases around the onset of plasma cell differentiation, with only a proportion of plasma cells containing CD79 (Comans-Bitter, 1997)(Nakamura, 1997). The HM57 antibody was clustered as Anti-CD79αcy at the Fifth International Workship on Human Leucocyte Differentiation Antigens (Engel, 1995).

Specificity

Antibody recognizes Human CD79αcy antigen.

Immunogen

Synthetic peptide sequence comprising amino acids 202-216 of the CD79α protein. This oligopeptide represents an intra-cytoplasmic C-terminal part of the protein.

Application

Milli-Mark Anti-CD79αcy-PE Antibody, clone HM57 is an antibody against CD79αcy for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by flow cytometry using human lymphocytes

Target description

82-95 kDa (Pilozzi,1998)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to PE in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintained refrigerated at 2-8°C from date of receipt. Protected from light in undiluted aliquots for 6 months from date of receipt.

Analysis Note

Control
Human Lymphocytes

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service