Skip to Content
Merck
All Photos(1)

Key Documents

EZFHEAD01

Millipore

Milliflex head for EZ-FIT® Filtration unit

suitable for (General microbiological analysis)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 18,600.00

CZK 18,600.00


In StockDetails


Request more information

Select a Size

Change View
1 EA
CZK 18,600.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

CZK 18,600.00


In StockDetails


Request more information

material

stainless steel

Quality Level

feature

ready-to-use

manufacturer/tradename

EZ-Fit®

input

sample type pharmaceutical(s)
sample type cosmetics
sample type beer
sample type bottled water

suitability

suitable for (General microbiological analysis)

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with EZ-Fit® System

shipped in

ambient

Application

General microbiological analysis

Preparation Note

Sterilization Method
Autoclavable

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX RACK&STACK is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service