Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Estimated to ship on20 July 2026fromAreál Kühne+Nagel spol. s r.o. | 17 600,00 CZK |
About This Item
species reactivity
human
Quality Segment
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
shipped in
dry ice
General description
Integrins belong to a large family of cell surface proteins commonly referred to as glycoproteins. Integrins are composed of noncovalently associated alpha and beta subunits which make up heterodimers: the larger termed alpha chain and the smaller beta chain. At least 16 alpha subunits and 8 beta subunits are expressed, which can give rise to 22 different integrins. Integrins are thought to play critical roles in cell migration, differentiation, and survival. There is now considerable evidence that these transmembrane receptors play an important role in the regulation of gene expression and signal transduction. Excellent reviews of integrin structure, function, and ligand specificity may be found in: Hynes, R. 1992. Cell 69:11-25; Kramer, R. 1993. "Integrin Structure and Ligand Specificity in Cell-Matrix Interactions" in Molecular and Cellular Aspects of Basement Membranes. Academic Press.
Chemicon offers the Collagen-Specific Integrins Investigator Kit for studies of collagen/integrin interactions. All kit reagents are also available in larger sizes. The reagents in this kit are not application specific but rather may be used in a variety of procedures, depending upon the needs of the individual investigator.
Physical form
Preparation Note
Important Note: Due to the small volumes contained within the kit, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge before opening to dislodge any liquid entrapped in the container′s cap during shipment.
Other Notes
alpha2 Mouse Monoclonal Antibody (Part No. MAB1950Z-20)- 1 tube, 20 μg.
alpha3 Mouse Monoclonal Antibody (Part No. MAB2056-20) - 1 tube, 20 μg.
alphaV Mouse Monoclonal Antibody (Part No. MAB1953Z-20)- 1 tube, 20 μg.
beta1 Mouse Monoclonal Antibody (Part No. MAB1951Z-20) - 1 tube, 20 μg.
beta3 Monoclonal Antibody (Part No. MAB1957Z-20) - 1 tube, 20 μg.
alphaVbeta3 Mouse Monoclonal Antibody (Part No. MAB1976Z-20)- 1 tube, 20 μg.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) ELISA: suitable | technique(s) ELISA: suitable, immunohistochemistry: suitable | technique(s) ELISA: suitable, flow cytometry: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable | technique(s) ELISA: suitable, flow cytometry: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable |
| manufacturer/tradename Chemicon® | manufacturer/tradename Chemicon® | manufacturer/tradename Chemicon® | manufacturer/tradename Chemicon® |
| shipped in dry ice | shipped in - | shipped in dry ice | shipped in dry ice |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
Storage Class
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



