Skip to Content
Merck

Skip To

DR1015

Anti-RENT1 Goat pAb

liquid, Calbiochem®

Promotion Available

Synonym(s):

Anti-HUPF1, Anti-NORF1

Sign In to View Organizational & Contract Pricing.

Spend 10 750 CZK — Save 5%; Spend 15 250 CZK — Save 10%; Spend 28 000 CZK — Save 15%*

Sign in to check eligibility. Shop Products

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship on01 October 2026fromAreál Kühne+Nagel spol. s r.o.

11 200,00 CZK

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Clone:
polyclonal
Species reactivity:
human
Application:
Citations:
-

11 200,00 CZK

Save Up To 15%


Estimated to ship on01 October 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

dilution

(Immunoblotting (1:1000-1:2500))

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... UPF1(5976)

General description

This Anti-RENT1 Goat pAb is validated for use in Immunoblotting for the detection of RENT1.
Immunoaffinity purified goat polyclonal antibody. Recognizes the ~140 kDa RENT1 protein.
Recognizes the ~140 kDa RENT1 protein in HeLa cells.

Immunogen

a synthetic peptide corresponding to amino acids at the C-terminus of human RENT1

Application


Immunoblotting (1:1000-1:2500)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In Tris/citrate phosphate buffer, pH 7.0-8.0.

Other Notes

May detect an additional band at ~80 kDa. Antibody should be titrated for optimal results in individual systems.
Mendell, J.T., et al. 2002. Science298, 419.
Medghalachi, S.M., et al. 2001. Hum. Mol. Genet.10, 99.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Highly Toxic (H)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PLA0010AP1063324870
description

liquid, Calbiochem®

description

Powered by Bethyl Laboratories, Inc.

description

liquid, Calbiochem®

description

liquid, Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

goat

biological source

goat

biological source

rabbit

biological source

rabbit

form

liquid

form

-

form

liquid

form

liquid

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

species reactivity

human

species reactivity

human, mouse

species reactivity

mouse, human, rat

species reactivity

human, rat


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters