Skip to Content
Merck

Skip To

DAWP14250

MCE Membrane Filter, 0.65 μm Pore Size

MF-Millipore, filter diam. 142 mm, hydrophilic

Synonym(s):

MF-Millipore Membrane Filter, 0.65 µm pore size

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 filters

Estimated to ship on07 July 2026fromAreál Kühne+Nagel spol. s r.o.

12 400,00 CZK

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
0.65 μm pore size, 81 % porosity
Material:
mixed cellulose esters (MCE) membrane, plain filter, white filter
Feature:
hydrophilic
Diam.:
142 mm
Sterility:
non-sterile

12 400,00 CZK


Estimated to ship on07 July 2026Details


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

description

142 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 50 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore, Millipore

parameter

140 mL/min-cm2 water flow rate, 75 °C max. temp., 9 L/min-cm2 air flow rate

diam.

142 mm

filter diam.

142 mm

thickness

150 μm

gravimetric extractables

3%

refractive index

n/D 1.51

matrix

MF-Millipore

pore size

0.65 μm pore size, 81 % porosity

capacity

150 μg/cm2 binding capacity (protein)

bubble point

≥1.17 bar, air with water at 23 °C

shipped in

ambient

General description

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.

Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Filter Code: DAWP

Application

  • Particle removal
  • Dairy microbiology
  • Retention of yeasts, molds, and algae
  • Microplastics

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: General laboratory filtration
  • Intended Use: Retention or removal of biological contaminants
  • Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
  • Storage Statement: Store in dry location away from heat source
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
DAWP09025DAWP01300DAWP04700
pore size

0.65 μm pore size, 81 % porosity

pore size

0.65 μm pore size, 81 % porosity

pore size

0.65 μm pore size, 81 % porosity

pore size

0.65 μm pore size, 81 % porosity

parameter

140 mL/min-cm2 water flow rate, 9 L/min-cm2 air flow rate, 75 °C max. temp.

parameter

140 mL/min-cm2 water flow rate, 75 °C max. temp., 9 L/min-cm2 air flow rate

parameter

140 mL/min-cm2 water flow rate, 75 °C max. temp., 9 L/min-cm2 air flow rate

parameter

140 mL/min-cm2 water flow rate, 75 °C max. temp., 9 L/min-cm2 air flow rate

manufacturer/tradename

MF-Millipore

manufacturer/tradename

MF-Millipore

manufacturer/tradename

MF-Millipore

manufacturer/tradename

MF-Millipore

material

mixed cellulose esters (MCE) membrane, white filter, plain filter

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

sterility

non-sterile

sterility

non-sterile

sterility

-

sterility

non-sterile

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic


Still not finding the right product?


signalword

Danger

Storage Class

4.1B - Flammable solid hazardous materials

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - ED ENV 1 - Eye Irrit. 2 - Flam. Sol. 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

9036-19-5

CAS No.

9036-19-5

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters