CFMQ18501
Photooxidation UV Lamp
For use with Milli-Q® Plus (1989-1996) systems
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
JA.14
Recommended Products
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® Plus
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
Replacement UV Lamp for photooxidation of organic compounds, used with Milli-Q® and Milli-Q® Plus (1989-1996) systems.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.


Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service