CC054-M
Human Collagen Type III
Synonym(s):
alpha 1 Type III Collagen
About This Item
Recommended Products
biological source
human
Quality Level
Assay
90% (SDS-PAGE)
form
liquid
manufacturer/tradename
Chemicon®
technique(s)
cell culture | mammalian: suitable
impurities
<1% human collagen types II, IV-VI, and non-collagen proteins
<10% human collagen type I
input
sample type: human embryonic stem cell(s)
sample type pancreatic stem cell(s)
sample type mesenchymal stem cell(s)
sample type neural stem cell(s)
sample type hematopoietic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type epithelial cells
NCBI accession no.
UniProt accession no.
General description
Physical form
Storage and Stability
Analysis Note
Legal Information
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service