Skip to Content
Merck

Skip To

CBL304

Anti-Serum Amyloid P Antibody, clone 5.4A

clone 5.4A, Chemicon®, from mouse

Synonym(s):

Component P

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
CZK 9,460.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
5.4A, monoclonal
Application:
ELISA
Citations:
2

CZK 9,460.00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

5.4A, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... APCS(325)

General description

Amyloid consists mainly of rigid, non-branching protein fibrils, together with rod-like aggregates of a pentagonal-shaped glycoprotein called amyloid P protein. Amyloid P protein, also known as component P, comprises 10 per cent of amyloid tissue and is present in all but the central nervous system forms of amyloid. Amyloid P protein is a constituent of normal basement membranes and the microfibrillaryelastic fiber network.

Immunogen

Human serum amyloid P

Application

Anti-Serum Amyloid P Antibody, clone 5.4A detects level of Serum Amyloid P & has been published & validated for use in ELISA.
Development of ELISA assay for the estimation of human serum amyloid P using CBL 305 biotin conjugate as the labelled pair

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Biochem/physiol Actions

This antibody is specific for human serum amyloid P.

FUSION PARTNER: NS1 myeloma cell line

Physical form

Format: Purified
The monoclonal is presented at a concentration of 100ug in phosphate buffered saline containing 10mM sodium azide. We recommend that each laboratory determine an optimum working titre for use in its particular application.

Preparation Note

For use within 1 month of purchase store at +4°C, for long term storage aliquot antibody into small volumes and store at -20°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CBL305AB2286MABN12
Gene Information

human ... APCS(325)

Gene Information

human ... APCS(325)

Gene Information

human ... APP(351)
mouse ... App(11820)

Gene Information

human ... APP(351)

antibody form

affinity isolated antibody

antibody form

purified antibody

antibody form

serum

antibody form

purified immunoglobulin

biological source

mouse

biological source

mouse

biological source

rabbit

biological source

mouse

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

conjugate

-

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human, mouse

clone

5.4A, monoclonal

clone

5.4D.3B, monoclonal

clone

polyclonal

clone

G2-11, monoclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters