Skip to Content
Merck
All Photos(1)

Documents

BW-10X5ML-B

Millipore

Anti-CDE (clone MS-24/MS-201/MS-26/MS-80) Blood Grouping Reagent

BIOSCOT Anti-CDE from the clone MS-24/MS-201/MS-26/MS-80, Human IgM/IgG, Blood Grouping Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116102

biological source

human

Quality Level

clone

MS-24, MS-201, MS-80 Human IgM/MS-26 Human IgG, monoclonal

packaging

pkg of 10 × 5 mL

manufacturer/tradename

BIOSCOT

storage condition

do not freeze

concentration

70-90 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

7.1-7.3

isotype

IgG
IgM

application(s)

clinical testing

shipped in

ambient

storage temp.

2-8°C

General description

Contains 0.1% (w/v) Sodium Azide

Filtered through a 0.22μm terminal filter

Shelf Life: 36 months from date of filtration

Product formulation conforms to the requirements of the EU IVD directive.

For In Vitro use only.

Application

For use by Tube;Slide;Microplate;Gel and Column techniques
Vialled, Labeled

Legal Information

BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service