Skip to Content
Merck

Skip To

BI-2ML-B

Blood typing BIOSCOT IVD Reagent Other Specificities & AHG

BIOSCOT Anti-Jka from the clone MS-15, Human IgM, Blood Grouping Reagent

Synonym(s):

Anti-Jka (clone MS-15) Blood Grouping Reagent

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41116102
NACRES:
NA.47
Note: For further information on price, availability or to place an order, please contact us


biological source

human

Quality Segment

clone

MS-15, monoclonal

packaging

pkg of 2 mL

manufacturer/tradename

BIOSCOT

storage condition

do not freeze

concentration

30-55 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable, blood typing: suitable

pH

7.1-7.3

isotype

IgM

application(s)

clinical testing

shipped in

ambient

storage temp.

2-8°C

General description

Contains 0.1% (w/v) Sodium Azide

Filtered through a 0.22μm terminal filter

Shelf Life: 36 months from date of filtration

CE marked under the requirements of EU regulation 2017/746




For In Vitro use only .

Application

Tube technique
Vialled, Labeled

Legal Information

BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library