Skip to Content
Merck
All Photos(1)

Key Documents

BI0837

Sigma-Aldrich

Deblocking Reagent 3% Trichloroacetic Acid in Toluene (w/v)

Synonym(s):

dna

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006
Pricing and availability is not currently available.

product line

Novabiochem®

Quality Level

form

liquid

reaction suitability

reaction type: Acetylations
reagent type: Lewis acid

transition temp

flash point 4.4 °C

shipped in

ambient

storage temp.

room temp

Analysis Note

Color (APHA): 20 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Trichloroacetic acid: 2.95-3.05% w/v
Water (H2O): 500ppm max

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F

Flash Point(C)

4.4 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service