Skip to Content
Merck
All Photos(1)

Key Documents

B451-500S

Tryptic Soy Broth Ready-to-use Bottles

Lecithin, Tween®, bottle capacity 1 L, bottle filling volume 500 mL, closure type, white screw cap with septum and protector, box of 4 bottles

Synonym(s):

CASO Broth, Casein Soyabean Digest Broth, TSB, TSB with neutralizer

Sign Into View Organizational & Contract Pricing

Select a Size

4 X 500 ML
CZK 874.00

CZK 874.00


Estimated to ship onMay 15, 2025Details


Request more information

Select a Size

Change View
4 X 500 ML
CZK 874.00

About This Item

UNSPSC Code:
41106214

CZK 874.00


Estimated to ship onMay 15, 2025Details


Request more information

material

polycarbonate bottle

Quality Level

sterility

sterile; autoclaved

form

powder

feature

closure type white screw cap with septum and protector

composition

Lecithin
Tween®

packaging

box of 4 bottles

technique(s)

microbiological culture: suitable
sterility testing: suitable

bottle capacity

1 L

bottle filling volume

500 mL

capacity

1000 mL

pH

7.3±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical

storage temp.

2-25°C

suitability

nonselective for bacteria

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tryptic Soy Broth (TSB) with Lecithin and Polysorbate 20(Tween®20) is a complex medium designed for the enumeration of total microbial count present in the test sample. Lecithin along with Polysorbate 80 is effective against quaternary ammonium compounds, amphoteric surfactants, benzamidines, chlorhexidines, and dequadin. Polysorbate 80 inactivates benzyl alcohol, carbanilides, di-chlorobenzyl alcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols, phenylethyl alcohols, and Solbrol (PHB esters).
Upon incubation, the enumerated microflora can be plated, isolated, and identified.

Application

Tryptic Soy Broth (TSB) with Lecithin and Polysorbate 20(Tween®20) is a complex medium used to enumerate microorganisms present in sterile or non-sterile products from food or pharmaceutical industry

Legal Information

TWEEN is a registered trademark of Croda International PLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service