Skip to Content
Merck

Skip To

APFA09050

Glass Fiber Membrane Filter, 1.6 μm Pore Size

Millipore, filter diam. 90 mm, hydrophilic

Synonym(s):

Glass Fiber Filter without binder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 filters

Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

1 750,00 CZK

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
1.6 μm pore size, 90 % porosity
Material:
glass fiber membrane, plain filter, white filter
Feature:
hydrophilic
Diam.:
90 mm
Sterility:
non-sterile

1 750,00 CZK


Estimated to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

glass fiber membrane, plain filter, white filter

description

1.6 µm pore size, hydrophilic glass fiber, 90 mm diameter

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Millipore

parameter

5 mL/min-cm2 water flow rate, 500 °C max. temp.

diam.

90 mm

filter diam.

90 mm

thickness

1 mm

gravimetric extractables

300%

pore size

1.6 μm pore size, 90 % porosity

shipped in

ambient

General description

Filter Code: APFA
Filter Type: Depth filter
Overview
Millipore fibrous disc filters are available in a wide range of flow rates and throughput capacities. Our Glass Fiber filters are available with or without binder resins and can be sterilized by EO, gamma, or autoclave (121 °C at 1 bar).

Glass Fiber Filters with Binder Resin:
Millipore filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated liquids. They are also widely used for clarification of aqueous solutions.

Type AP15
•Lowest dirt-holding capacity
•Recommended prefilter for 0.2 to 0.6 µm filters

Type AP20
•Lower retention, higher dirt-holding capacity than AP15 filters
•Recommended prefilter for 0.8 to 8.0 µm filters
•Use upstream to protect type AP15 filters

Type AP25
•Added thickness yields same retention and greater dirt-holding capacity as AP20
•Recommended prefilter for 0.9 to 8 µm filters, especially for proteinaceous and heavily contaminated liquids
•Use upstream to protect type AP15 filters

Glass Fiber Filters without Binder Resin:
The filters without binder resin retain their structural integrity without weight loss when heated up to 500 °C and can therefore be used in gravimetric analysis as well as for the filtration of hot gases.

Type APFA
•Retains fine particles with good efficiency, even at high flow rates
•Recommended for monitoring wastewater and collecting suspended particles in gases (e.g., fumes from chimneys)
•Use in biochemical applications (e.g., collection of cells and filtration of protein or nucleic acid precipitates)

Type APFB
•Higher mechanical strength when wet and greater loading capacity than type APFA filters
•Use for liquid clarification, quantification of solids in suspensions of fine particles, and scintillation counting

Type APFC
•Greater retention, especially for fine particulate and microorganism removal than type APFA filters
•Use for determining total suspended solids in drinking water
•Use for filtering proteins or nucleic acid TCA precipitates and for collecting cells and microorganisms

Type APFD
•Thick filter with high flow and low retention
•Use for clarifying suspensions containing particulates greater than 1.0 µm

Type APFF
•Use for filtering extremely fine precipitates such as protein, nucleic acids, or serum precipitates
•Recommended for USEPA method 1311 for TCLP analysis

Type AP40
•Recommended for Standard Methods Total Suspended Solids 2540D
•Recommended for USEPA method 1311 for TCLP analysis
•Maintains structural integrity without weight loss when ignited to 550 °C (1022 °F) after sample filtration
•Recommended for determining volatile suspended matter in wastewater and industrial effluents

Application

  • Fine particle retention
  • Monitoring wastewater
  • Collecting suspended particles in gases
  • Collection of cells
  • Filtration of protein or nucleic acid precipitates


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
APFA04700APFF02500APFF04700
parameter

5 mL/min-cm2 water flow rate, 500 °C max. temp.

parameter

5 mL/min-cm2 water flow rate, 500 °C max. temp.

parameter

1.4 mL/min-cm2 water flow rate, 500 °C max. temp.

parameter

1.4 mL/min-cm2 water flow rate, 500 °C max. temp.

pore size

1.6 μm pore size, 90 % porosity

pore size

1.6 μm pore size, 90 % porosity

pore size

0.7 μm pore size, 90 % porosity

pore size

0.7 μm pore size, 90 % porosity

manufacturer/tradename

Millipore

manufacturer/tradename

Millipore

manufacturer/tradename

Millipore

manufacturer/tradename

Millipore

diam.

90 mm

diam.

47 mm

diam.

25 mm

diam.

-

material

glass fiber membrane, white filter, plain filter

material

glass fiber membrane, plain filter, white filter

material

glass fiber membrane, plain filter, white filter

material

glass fiber membrane, plain filter, white filter

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile


Still not finding the right product?


Storage Class

10-13 - German Storage Class 10 to 13



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters