AG222
Dopamine D4 Receptor, control peptide for AB1789/AB1789P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.25
concentration
0.2 mg/mL
Quality Level
technique(s)
cell based assay: suitable
UniProt accession no.
Gene Information
rat ... DRD4(25432)
General description
FUNCTION: SwissProt: P21917 # This is one of the five types (D1 to D5) of receptors for dopamine. The activity of this receptor is mediated by G proteins which inhibit adenylyl cyclase.
SIZE: 467 amino acids; 48361 Da
SUBUNIT: Interacts with CLIC6 (By similarity) and GPRASP1.
SUBCELLULAR LOCATION: Cell membrane; Multi-pass membrane protein.
SIMILARITY: SwissProt: P21917 ## Belongs to the G-protein coupled receptor 1 family.
SIZE: 467 amino acids; 48361 Da
SUBUNIT: Interacts with CLIC6 (By similarity) and GPRASP1.
SUBCELLULAR LOCATION: Cell membrane; Multi-pass membrane protein.
SIMILARITY: SwissProt: P21917 ## Belongs to the G-protein coupled receptor 1 family.
Application
For use in blocking the staining of AB1789 or AB1789P.
Optimal working dilution must be determined by the end user.
Optimal working dilution must be determined by the end user.
Physical form
Liquid in PBS, pH 7.4 containing 0.1% sodium azide.
Storage and Stability
Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service