Skip to Content
Merck

Skip To

ABT52

Anti-phospho-Peak Antibody (Tyr665)

from rabbit, purified by affinity chromatography

Synonym(s):

Pseudopodium-enriched atypical kinase 1, Sugen kinase 269, Tyrosine-protein kinase SgK269

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship onJune 05, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 11,100.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human
Application:
DB, WB
Citations:
2

CZK 11,100.00


Estimated to ship onJune 05, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), rat (based on 100% sequence homology), orangutan (Sumatran; based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

dot blot: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr665)

Gene Information

human ... PEAK1(79834)

General description

220 kDa calculated
Pseudopodium-enriched atypical kinase 1 (Peak1; KIAA2002, or sgk269) is a ubiquitous non-receptor tyrosine kinase that belongs to the NFK3 family. It interacts with actin filaments via its N-terminal actin-targeting region. It also modulates the activity of multiple cytoskeletal regulatory proteins such as paxillin, p130Cas, Erk, and Crk; and mediates cell proliferation and migration via the Ras/Raf/Erk and Src-p130Cas-Crk-Paxillin pathways. Peak1 may play an important role in the progression of tumors.

Immunogen

Epitope: Phosphorylated Tyr665
KLH-conjugated linear peptide corresponding to human Peak phosphorylated at Tyr665.

Application

This phospho-Peak antibody is validated for use in WB & Dot Blot for the detection of the phospho-Peak protein.
Western Blot Analysis: A representative lot was used by an independent laboratory in 293T cells transfected with wildtype GFP-Peak. (Dr. Richard Klemke, University of California-San Diego.)

Biochem/physiol Actions

This antibody recognizes Peak phosphorylated at Tyr665.

Analysis Note

Control
Modified and non modified peptides of phosphorylated and non-phosphorlyated Peak proteins
Evaluated by Dot Blot in modified and non modified peptides of phosphorylated and non-phosphorlyated Peak proteins.

Dot Blot (Specificity) Analysis: A representative lot blocked phospho-Peak (Tyr665) in modified and non modified peptides of phosphorylated and non-phosphorlyated Peak proteins.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABC212ABS92ABS219
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified antibody

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

technique(s)

dot blot: suitable, western blot: suitable

technique(s)

inhibition assay: suitable (peptide), western blot: suitable

technique(s)

western blot: suitable

technique(s)

western blot: suitable

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

UniProt accession no.

Q9H792

UniProt accession no.

O70405

UniProt accession no.

Q13470

UniProt accession no.

Q02763


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters