Skip to Content
Merck

Skip To

ABT256

Anti-Collagen Type I (COL1A1) Antibody

rabbit polyclonal

Synonym(s):

Collagen alpha-1(I) chain, Alpha-1 type I collagen

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 10,900.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Clone:
polyclonal
Species reactivity:
mouse, human, sheep
Application:
FACS, IHC, WB
Citations:
1

CZK 10,900.00


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-Telo-Collagen Type I, A1/COL1A1, from rabbit, purified by affinity chromatography

biological source

rabbit

Quality Segment

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, sheep

technique(s)

flow cytometry: suitable, immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... COL1A1(1277)

General description

Collagen alpha-1(I) chain (UniProt P02452; also known as Alpha-1 type I collagen) is encoded by the COL1A1 gene (Gene ID 1277) in human. Collagen is the major component of the extracellular matrix (ECM) and form the fibrils of tendons, ligaments, and bones. Type I collagen consists of two alpha I chains and one alpha 2 chain. Alpha-1 type I collagen is initially produced as an 1464-amino acid prepro-form with a signal peptide sequence (a.a. 1-22) and two propeptide sequences (a.a. 23-161 and a.a. 1219 –1464), the removal of which yields the mature alpha-1(I) chain. The mature alpha-1(I) chain is composed mostly of a large triple-helical region (a.a. 179-1192) sandwiched between two nonhelical segments known as the N-terminal telopeptide (a.a. 162-178; numbering based on the prepro-form) and the C-terminal telopeptide (a.a. 1193-1218; numbering based on the prepro-form). Collagen can be extracted from tissue via either enzymatic or non-enzymatic means. Collagen extracted using the proteolytic enzyme pepsin corresponds to the large triple-helical region, referred to as atelocollagen because both the N- and C-terminal telopeptides have been cleaved off by pepsin. On the other hand, collagen preparations obtained with non-enzymatic means (e.g. by acid extraction) have the intact telopeptides at both ends.
~180 kDa observed

Immunogen

Epitope: C-terminal telopeptide region
Linear peptide corresponding to sequence derived from human Collagen Type I, A1/COL1A1 C-terminal telopeptide region.

Application

Anti-Telo-Collagen Type I Antibody, A1/COL1A1 is an antibody against Telo-Collagen Type I for use in Western Blotting, Flow Cytometry, Immunohistochemistry.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
Western Blotting Analysis: A representative lot detected Telo-Collagen Type I, A1/COL1A1 in mouse lung and skin tissue lysate.
Flow Cytometry Analysis: A representative lot detected Telo-Collagen Type I, A1/COL1A1 in mouse lung tissue.
Immunohistochemistry Analysis: A representative lot detected Telo-Collagen Type I, A1/COL1A1 in mouse lung tissue.

Biochem/physiol Actions

Cat. No. ABT256 Anti-Telo-Collagen Type I, A1/COL1A1 was raised against a sequence derived from the C-terminal telopeptide (not the N-terminal telopeptide) region. This polyclonal antibody will detect prepro-, pro-, and mature collagen alpha-1(I) chain, but not atelocollagen prepared with proteolytic enzymes due to the absence of the C-terminal telopeptide sequence.
Predicted to react with a broad range of species based on high sequence homology, including mammalian, avian, and amphibian species.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in mouse liver tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Telo-Collagen Type I, A1/COL1A1 in 1 µg of mouse liver tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABT257ABT123HPA011795
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

UniProt accession no.

P02452

UniProt accession no.

P02452

UniProt accession no.

P02452

UniProt accession no.

P02452

species reactivity

mouse, human, sheep

species reactivity

mouse, human

species reactivity

mouse, rat

species reactivity

human

technique(s)

flow cytometry: suitable, western blot: suitable, immunohistochemistry: suitable

technique(s)

flow cytometry: suitable, immunohistochemistry: suitable, western blot: suitable

technique(s)

ELISA: suitable, immunohistochemistry: suitable (paraffin), western blot: suitable

technique(s)

immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:50-1:200


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters