Select a Size
CZK 11,900.00
CZK 11,900.00
Estimated to ship onMarch 18, 2026Details
About This Item
Skip To
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Quality Level
Gene Information
human ... CH25H(9023)
1 of 4
This Item | |||
|---|---|---|---|
| antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| Quality Level 100 | Quality Level 100 | Quality Level - | Quality Level - |
| biological source rabbit | biological source mouse | biological source rabbit | biological source rabbit |
| shipped in wet ice | shipped in dry ice | shipped in wet ice | shipped in wet ice |
| species reactivity human | species reactivity human | species reactivity - | species reactivity - |
| UniProt accession no. | UniProt accession no. | UniProt accession no. - | UniProt accession no. - |
General description
Immunogen
Application
Signaling
Signaling Neuroscience
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: 2.0 µg/mL of this antibody detected CH25H in 10 µg of THP-1 cell lysate.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


