Skip to Content
Merck
All Photos(2)

Key Documents

ABS551

Sigma-Aldrich

Anti-Glucagon receptor Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

GL-R, Glucagon receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 10,500.00

CZK 10,500.00


Estimated to ship onApril 11, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 10,500.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 10,500.00


Estimated to ship onApril 11, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

primate (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GCGR(2642)

General description

Glucagon receptor or GL-R (UniProt P47871) is encoded by the GCGR (also known as GGR) gene (Gene ID 2642) in human. Glucagon is a 29-amino acid peptide that stimulates glycogenolysis and gluconeogenesis in the liver to increase blood glucose. The receptor for glucagon is a class 2 (or class B) G protein-coupled receptor (GPCR) that signals through Gs to stimulate cAMP production. Mice lacking the glucagon receptor have mild hypoglycemia after fasting and exhibit hyperplasia of pancreatic alpha-cells. Because of its role in promoting hyperglycemia, the glucagon receptor presents a potential target for treatment of diabetes.

Immunogen

Epitope: Fourth cytoplasmic domain
KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human Glucagon receptor.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Glucagon receptor in human pancreas.
This Anti-Glucagon receptor Antibody is validated for use in Western Blotting, Immunohistochemistry (Paraffin) for the detection of Glucagon receptor.

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Glucagon receptor in 10 µg of human liver tissue lysate.

Target description

~59 kDa observed. Target band appears larger than the calculated molecular weight of 54.0 kDa (Preform) and 51.3 kDa (mature) due to glycosylation. Lower molecular weight degradation band(s) can also be seen (Iwanij, V., et al. (1990). J. Biol Chem. 265(34):21302-21308).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service