Skip to Content
Merck
All Photos(1)

Key Documents

ABS224

Sigma-Aldrich

Anti-YVH1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Dual specificity protein phosphatase 12, Dual specificity tyrosine phosphatase YVH1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 9,450.00

CZK 9,450.00


Estimated to ship onApril 03, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
CZK 9,450.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 9,450.00


Estimated to ship onApril 03, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DUSP12(11266)

General description

Yeast VH1-related phosphatase (YVH1), also known as “dual specificity protein phosphatase 12” (DUSP12) is a ubiquitous phosphatase, with relatively high levels of expression in the ovary, testis, spleen, and peripheral blood leukocytes. YVH1 primarily removes phosphate groups from phosphorylated serine, threonine, and tyrosine residues in its target substrates. However, YVH1 also contains a zinc-binding domain which may enable it to interact with other proteins involved in cell growth, cell survival, and cell cycle progression. YVH1 may also be involved in the assembly of ribosomes, and in the biogenesis of the 60S ribosomal subunit. It can be targeted to different subcellular locations by phosphorylation of its Ser335 residue located in the zinc-binding domain.

Immunogen

GST-tagged recombinant protein corresponding to human YVH1.

Application

Anti-YVH1 Antibody is an antibody against YVH1 for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Kinases & Phosphatases
Western Blot Analysis: A representative lot detected YVH1 in 10 µg of Jurkat cell lysate.

Quality

Evaluated by Western Blot in HeLa cells.

Western Blot Analysis: A 1:500 dilution of this antibody detected YVH1 in 10 µg of HeLa cell lysate.

Target description

~38 kDa observed. An uncharacterized band at ~36 kDa may be observed in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cells

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service