ABS2124
Anti-Ferrochelatase
serum, from rabbit
Synonym(s):
Heme synthase, protoheme ferro-lyase
About This Item
biological source
rabbit
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human, mouse
packaging
antibody small pack of 25 μL
technique(s)
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... FECH(2235)
General description
Specificity
Immunogen
Application
Western Blotting Analysis: A representative lot detected Ferrochelatase in differentiated mouse erythroleukemia (MEL) cells and transfected heterologous cells. (Chen, W., et. al. (2010). Blood. 116(4):628-30; Anand, S., et. al. (2009). Clin Cancer Res. 15(10):3333-43; Medlock, A.E., et. al. (2015). PLoS One. 10(8):e0135896).
Quality
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Ferrochelatase in 10 µg of K562 cell lysate.
Target description
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service