Skip to Content
Merck

Skip To

ABS21

Anti-Surfactant Protein B Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Pulmonary surfactant-associated protein B, 18 kDa pulmonary-surfactant protein, Pulmonary surfactant-associated proteolipid SPL(Phe), SP-B

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL

Estimated to ship onJune 04, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 10,500.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
1

CZK 10,500.00


Estimated to ship onJune 04, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SFTPB(6439)

General description

Surfactant Protein B (SP-B) is mostly composed of phospholipids, but also incorporates essential proteins. It is necessary for pulmonary surfactant function and the alveolar type 2 cell phenotype. SP-B deficiency has been known to cause congenital pulmonary alveolar proteinosis. SP-B is thought to protect lungs against endotoxin-induced inflammation. SP-B could be used as a clinical marker in chronic heart failure.
~16 kDa observed

Immunogen

Epitope: Saposin B-type 2 domain
GST-tagged recombinant protein corresponding to human Surfactant Protein B at the Saposin B-type 2 domain.

Application

Immunohistochemistry Analysis: 1:200 dilution from a representative lot detected Surfactant protein B in clara cells in normal rat lung tissue.
Research Category
Signaling
Research Sub Category
Lipid Signaling
This Anti-Surfactant Protein B Antibody is validated for use in WB, IH(P) for the detection of Surfactant Protein B.

Biochem/physiol Actions

This antibody recognizes Surfactant Protein B at the Saposin B-type 2 domain.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human lung tissue lysate
Evaluated by Western Blot in human lung tissue lysate.

Western Blot Analysis: 1:5,000 dilution of this antibody detected Surfactant Protein B on 10 µg of human lung tissue lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB2102128HPA062148ABC99
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

species reactivity

rat, mouse, human

species reactivity

dog, guinea pig, human, rat

species reactivity

human

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

technique(s)

immunohistochemistry: suitable, western blot: suitable

technique(s)

immunohistochemistry: 1:1000- 1:2500

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable


Still not finding the right product?

Explore all of our products under Anti-Surfactant Protein B Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters