Skip to Content
Merck

ABS1587

Anti-Ribophorin I/RPN-I Antibody

serum, from rabbit

Synonym(s):

Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1, Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit, Ribophorin I, RPN-I, Ribophorin-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, canine, human

species reactivity (predicted by homology)

nonhuman primates (based on 100% sequence homology), porcine (based on 100% sequence homology), monkey (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... RPN1(6184)

General description

Dolichyl-diphosphooligosaccharide-protein glycosyltransferase subunit 1 (EC 2.4.99.18; UniProt P04843; also known as Dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit, Ribophorin I, RPN-I, Ribophorin-1) is encoded by the RPN1 gene (Gene ID 6184) in human. Ribophorin I is an ER transmembrane protein and a subunit of the mammalian Oligosaccharyltransferase (OST) complex. Ribophorin I binds newly synthesized target proteins and delivers them to the catalytic core of the OST complex. Association of ribophorin I with newly synthesized membrane proteins was observed after the exit of the nascent chain from the Sec61 translocon, and it was unaffected by the N-glycosylation status. Ribophorin I is reported to preferentially interact with misfolded over native ribonuclease A. In addition, Ribophorin I is shown to complex with another ER protein malectin and enhance malectin′s ability to prevent the secretion of the folding-defective proteins.
~68 kDa observed.

Immunogen

Epitope: C-terminus.
KLH-conjugated linear peptide corresponding to the C-terminal sequence of human Ribophorin I/RPN-I.

Application

Research Category
Signaling
Research Sub Category
Organelle & Cell Markers
This Anti-Ribophorin I/RPN-I Antibody is validated for use in Western Blotting for the detection of Ribophorin I.
Western Blotting Analysis: An 1:5,000 dilution from a representative lot detected ribophorin I in in 25 µg of HeLa cell lysate (Courtesy of Dr. Christopher V. Nicchitta, Duke University Medical Center, Durham, NC).
Western Blotting Analysis: A representative lot detected ribophorin I in the alkali-extracted and Triton X-114-selected integral membrane fraction derived from UV-cross-linked canine pancreas rough microsomes (RM) preparation (Jagannathan, S., et al. (2014). J. Biol. Chem. 2014, 289(37):25907-25924).
Western Blotting Analysis: A representative lot detected ribophorin I in both the Brij35-sensitive (BrS) and Brij35-resistant (BrR) fraction of HeLa ER membrfane proteins (Jagannathan, S., et al. (2014). J. Biol. Chem. 2014, 289(37):25907-25924).
Western Blotting Analysis: A representative lot detected ribophorin I associated with the rRNA-free poly(A) messenger ribonucleoproteins (mRNPs) purified by oligo(dT) from UV-cross-linked canine pancreas rough microsomes (RM) preparation (Jagannathan, S., et al. (2014). J. Biol. Chem. 2014, 289(37):25907-25924).
Western Blotting Analysis: A representative lot detected ribophorin I in both the smooth and rough microsomes sub-fractionated from a rat liver microsome preparation (Menon, A.K., et al. (2000). Curr. Biol. 10(5):241-252).
Western Blotting Analysis: A representative lot detected ribophorin I in canine rough microsome (RM) fractions obtained by sucrose gradient centrifugation of digitonin- or DHPC-solubilized canine RM preparation with or without ribosomes stripping by EDTA treatment (Potter, M.D., and Nicchitta, C.V. (2000). J. Biol. Chem. 275(3):2037-2045).

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in canine pancreas tissue rough microsome lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Ribophorin I/RPN-I in 2.5 µL of canine pancreas tissue rough microsome lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library