Skip to Content
Merck
All Photos(1)

Key Documents

ABS1531

Sigma-Aldrich

Anti-AGS3/GPSM1 Antibody

serum, from rabbit

Synonym(s):

G-protein-signaling modulator 1, Activator of G-protein signaling 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 9,870.00

CZK 9,870.00


Estimated to ship onMarch 21, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
CZK 9,870.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 9,870.00


Estimated to ship onMarch 21, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

G-protein-signaling modulator 1 (UniProt Q9R080; also known as Activator of G-protein signaling 3) is encoded by the Gpsm1 (also known as Ags3) gene (Gene ID 246254) in rat species. AGS3/GPSM1 is an evolutionarily conserved protein with four G-protein regulatory (GPR) GoLoco motifs (a.a. 493-515, 546-568, 594-616, 628-650 of murine GPSM1) and functions as a guanine nucleotide dissociation inhibitor (GDI). GPSM1 increases heteromeric polycystin-1/polycystin-2 ion channel activity via Gβγ subunits. GPSM1 is involved in regulating mitotic spindle orientation, cell polarity, and adenylyl cyclase activity. GPSM1 plays a role in renal epithelial cell repair following renal injury and GPSM1 induction in polycystic kidney disease (PKD) and may be a critical modulator of the renal cystogenic process.

Specificity

This polyclonal antiserum immunostained predominantly renal collecting ducts of postnatal P12 ADPKD mice. Immunogen peptide blocking eliminated antibody stianing (Kwon, M., et al. (2012). Proc. Natl. Acad. Sci. U. S. A. 109(52):21462-21467). This antiserum targets a STK11/LKB1-interacting region sequence present in rat AGS3/GPSM1 isoform 1 and isoform 4 (isoform Long), 28% present in isoform 3 (Short-2), but completely absent from isoforms 2 (Short-1) reported by UniProt (Q9R080). Target sequence is also present in mouse isoforms 1, 3, 4, and 5, but absent from isoform 2 ( (UniProt Q6IR34).

Immunogen

Epitope: STK11/LKB1-interacting region.
Linear peptide corresponding to a sequence within the STK11/LKB1-interacting region of rat AGS3/GPSM1.

Application

Immunofluorescence Analysis: A representative lot immunostained predominantly renal collecting ducts by fluorescent immunohistochemistry using kidney sections from postnatal P12 Pkd1v/v ADPKD mice (Kwon, M., et al. (2012). Proc. Natl. Acad. Sci. U. S. A. 109(52):21462-21467).
Immunohistochemistry Analysis: A representative lot detected a time-dependent induction of AGS3/GPSM1 immunoreactivity co-localized with that of the proliferation marker Ki-67 in formalin-fixed, paraffin-embedded kidney sections from rats following ischemia-reperfusion injury (IRI) initiated by a 30-min occlusion of renal blood flow via bilateral renal artery clamping (Regner, K.R., et al. (2011). FASEB J. 25(6):1844-1855).
Western Blotting Analysis: A representative lot detected developmental stage-dependent renal AGS3/GPSM1 expression level. High level was seen in E13.5 embryonic kidney and greatly reduced among postnatal mice with low-to-undetectable levels around postnatal day P28. Postnatal renal AGS3/GPSM1 levels were found elevated and remained detectable up to P28 among Pkd1v/v autosomal dominant polycystic kidney disease (ADPKD) mice expressing noncleavable polycystin-1/PC1 (Kwon, M., et al. (2012). Proc. Natl. Acad. Sci. U. S. A. 109(52):21462-21467).
Western Blotting Analysis: A representative lot detected much higher AGS3/GPSM1 expression level in brain than kidney from postnatal day 19 (P19) wild-type or Pkd1v/v ADPKD mice (Kwon, M., et al. (2012). Proc. Natl. Acad. Sci. U. S. A. 109(52):21462-21467).
Western Blotting Analysis: A representative lot detected a time-dependent renal AGS3/GPSM1 induction in rats following ischemia-reperfusion injury (IRI) initiated by a 40-min occlusion of renal blood flow (Regner, K.R., et al. (2011). FASEB J. 25(6):1844-1855).
Research Category
Signaling
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling
This Anti-AGS3 Antibody is validated for use in Western Blotting, Immunofluorescence, Immunohistochemistry (Paraffin) for the detection of GPSM1.

Quality

Evaluated by Western Blotting in embryonic mouse brain tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected AGS3/GPSM1 in 10 µg of E13-14 embryonic mouse brain tissue lysate.

Target description

~74/25 kDa oberved. 74.44 kDa (rat isoform 1), 72.05 kDa (rat isoform 4; isoform Long), and 25.08 kDa (rat isoform 3; isoform Short-2), 74.36/71.93/33.56/78.11 kDa (mouse isoform 1/3/4/5) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Rabbit polyclonal antibody serum with 0.05% sodium azide.
Unpurified.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Vural et al.
Journal of cell science, 133(17) (2020-08-02)
Activator of G-protein signaling 3 (AGS3, encoded by GPSM1) was discovered as a one of several receptor-independent activators of G-protein signaling, which are postulated to provide a platform for divergence between canonical and noncanonical G-protein signaling pathways. Similarly, Dishevelled (DVL)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service