Skip to Content
Merck
All Photos(1)

Key Documents

ABS1509

Sigma-Aldrich

Anti-phospho-Neph1 (Tyr637/638) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Kin of IRRE-like protein 1, Kin of irregular chiasm-like protein 1, Nephrin-like protein 1, phospho-Neph1 (Tyr637/638)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 9,740.00

CZK 9,740.00


Estimated to ship onApril 11, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
CZK 9,740.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 9,740.00


Estimated to ship onApril 11, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr637/pTyr638 )

Gene Information

human ... KIRREL1(55243)

General description

Kin of IRRE-like protein 1 (UniProt Q80W68; also known as Nephrin-like protein 1, Kin of IRRE like 1, Kin of irregular chiasm-like protein 1) is encoded by the Kirrel (also known as 6720469N11Rik, Kirrel1, Neph1) gene (Gene ID 170643) in murine species. Podocytes are specialized epithelial cells that are critical components of the glomerular filtration barrier. Glomerular injury, such as that caused by renal ischemia, is often characterized by the effacement of podocytes, loss of slit diaphragms, and proteinuria. The podocyte proteins Neph1 and nephrin are critical for maintaining the structural integrity of slit diaphragm and therefore the filtration function of the glomerulus by organizing a signaling complex at the podocyte cell membrane. The cytoplasmic domain of Neph1 plays a key role in actin cytoskeleton remodeling events that are directly associated with Fyn-mediated Neph1 pTyr637/638 phosphorylation that leads to the recruitment of adapter proteins and signaling molecules, such as Grb2, Csk tyrosine kinase, and ZO-1. Phosphorylation-induced association between Neph1 and ZO-1 plays a key role in defining the tight junction formation in podocytes.

Specificity

Mouse Neph1 Tyr637/638 site is equivalent to human Neph1 Tyr605/606. Equivalent target site is also present in human spliced isoforms 2&3 (Tyr621/622 & Tyr502/503, respectively), but not in murine spliced isoform 2.

Immunogen

Epitope: Cytoplasmic domain
Linear peptide corresponding to the cytoplasmic domain of mouse phospho-Neph1 (Tyr637/638).

Application

Detect Neph1 using this rabbit Polyclonal Anti-phospho-Neph1 Antibody (Tyr637/638), Cat. No. ABS1509, validated for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
Western Blotting Analysis: A representative lot detected Neph1 phosphorylation in kinase assay using Fyn and COS-7 lysate containing exogenously expressed Neph1 (Data courtesy of Deepak Nihalini, University of Pennsylvania)
Western Blotting Analysis: A representative lot detected Neph1 phosphorylation in lysates from puromycin aminonucleoside-/PAN-treated human pocodytes (Arif, E., et al. (2014). J Biol Chem. 289(14):9502-9518).

Quality

Evaluated by Western Blotting in pervanadate treated human podocyte cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Neph1 phosphorylation at the target tyrosine residues in 7.5 µL of pervanadate treated human podocyte cell lysate.

Target description

~120 kDa observed. Target band size appears larger than the calculated molecular weight (87.2 kDa/murine isoform 1, 72.4-85.0 kDa/human isoform 1-3) due to posttranslational modifications (glycosylation & phosphorylation). Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing PBS/glycine/5% BSA, .02% Sodium Azide and 30% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service