Skip to Content
Merck

Skip To

ABN249

Anti-HNE Michael Adducts Antibody, Reduced

serum, from rabbit

Synonym(s):

Reduced 4-hydroxy-trans-2-nonenal Michael adducts, Reduced HNE Michael adducts

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL

Estimated to ship onJune 08, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 10,200.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
bovine, human
Application:
WB
Citations:
3

CZK 10,200.00


Estimated to ship onJune 08, 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

bovine, human

technique(s)

western blot: suitable

shipped in

ambient

target post-translational modification

unmodified

General description

Reactive aldehydes generated during lipid peroxidation contribute to cell and tissue damage associated with oxidative stress. Alpha,beta-unsaturated carbonyl compounds such as 4-hydroxy-2-nonenal (HNE), 4-oxo-2-nonenal (ONE), and acrolein, generated from oxidation of polyunsaturated fatty acids (PUFAs), readily interact with nucleophilic groups on proteins, leading to intramolecular or intermolecular protein crosslinking. Two types of HNE modifications exist, the first type is formed by a 1:1 Michael adduct reaction with lysine amine, histine imidazole, or cysteine sulfhydryl, while the second type is produced by one HNE reacting simultaneously with two free amine groups from two N-alpha-acetyllysines (NALs), so-called Michael-Schiff base adduct. In the latter case, the initial nonfluorescent 2:1 lysine-HNE Michael adduct-Schiff base further undergoes oxidative cyclization to form a fluorescent 2-hydroxy-3-imino-1,2-dihydropyrrol derivative commonly referred to as HNE fluorophore. Reversibly formed HNE-Lys Michael adduct is not stable and reduction by sodium borohydride (NaBH4) is often employed to preserve Lys HNE mofidication in protein samples. Reductive stabilization by NaBH4 treatment is also used to prevent 4-HNE loss from 4-HNE-Cys Michael adduct by retro-Michael addition reaction (RMA) during collision-induced dissociation (CID) and electron transfer dissociation (ETD) MS/MS.
Variable depending on the size(s) of the HNE-modified protein(s). Uncharacterized bands may be observed in some lysate(s).

Immunogen

HNE-modifed KLH chemically reduced by sodium borohydride.

Application

Anti-HNE Michael Adducts Antibody, Reduced, Cat. No. ABN249, is a highly specific rabbit polyclonal antibody that targets the reduced form of 4-Hydroxy-2-noneal Michael adducts and has been tested in Western Blotting.
Application Notes:
1. ABN249 replaces Calbiochem 393207. ABN249 is expected to work in immunocytochemistry and immunohistochemistry. Please refer to the following 393207 citations as examples of sample preparations and detection in these two types of applications:

Usatyuk, P.V., et al. (2006). J. Biol. Chem. 281(46):35554-35566 (Immunocytochemistry).
McCormack, A.L., et al. (2005). J. Neurochem. 93(4):1030-1037 (Immunohistochemistry).

2. Anti-HNE Fluorophore, Cat. No. 393206, is also available for the detection of HNE Fluorophore by ELISA, Immunohistochemistry, and Western blotting.

3. To generate a positive control for Western blotting, treat a 2 mg/mL BSA solution in 10 mM KH2PO4, pH 7.4, with 0.1 to 0.5 mM HNE and incubate for 1 hr at 37°C. The reaction can be monitored by the decrease of HNE absorbance at 224 nm (extinction coefficient 13,750/M). Free HNE can be removed by gel filtration and dialysis following the chemical reaction.

HNE Michael adducts in protein solutions must be chemically reduced by sodium borohydride (NaBH4) prior to analysis using ABN1348. A freshly prepared 100 mM NaBH4 solution in 1 mM NaOH is added 1:9 (V:V) to the protein sample solution to a final NaBH4 concentration of 10 mM. The protein solution is kept at room temperature for 10 to 30 min prior to quenching with the addition of 5 to 10 molar equivalents of acetone.

Biochem/physiol Actions

Target modification is not species-specific.
This polyclonal antiserum detected HNE-modified BSA only after, but not before, reduction by sodium borohydride.

Analysis Note

Control
HNE-modifed BSA chemically reduced by sodium borohydride.
Evaluated by Western Blotting in Reduced HNE-BSA.

Western Blotting Analysis: A 1:10,000 of this antibody detected 10 ng of HNE-modified BSA only after, but not before, reduction by sodium borohydride.

Other Notes

Concentration: Please refer to lot specific datasheet.
Replaces: Calbiochem 393207

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB5605ABT171ABN2268M
biological source

rabbit

biological source

goat

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

serum

antibody form

serum

antibody form

serum

antibody form

serum

shipped in

ambient

shipped in

dry ice

shipped in

wet ice

shipped in

ambient

species reactivity

bovine, human

species reactivity

human

species reactivity

monkey, mouse

species reactivity

rat, mouse, human

technique(s)

western blot: suitable

technique(s)

ELISA: suitable, western blot: suitable

technique(s)

ELISA: suitable, immunocytochemistry: suitable, western blot: suitable

technique(s)

immunocytochemistry: suitable, immunofluorescence: suitable, western blot: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters