Skip to Content
Merck
All Photos(1)

Key Documents

ABN1676M

Sigma-Aldrich

Anti-Calcipressin-1

serum, from rabbit

Synonym(s):

RCAN-1, Down syndrome critical region protein 1 homolog, Myocyte-enriched calcineurin-interacting protein 1, MCIP1, Regulator of calcineurin 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
CZK 3,890.00
100 μL
CZK 9,810.00

CZK 3,890.00


Estimated to ship onMarch 07, 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
CZK 3,890.00
100 μL
CZK 9,810.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 3,890.00


Estimated to ship onMarch 07, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Calcipressin-1 (Uniprot: Q6IN33; also known as Down syndrome critical region protein 1 homolog, Myocyte-enriched calcineurin-interacting protein 1, MCIP1, Regulator of calcineurin 1, RCAN-1) is encoded by the Rcan1 (also known as Dscr1) gene (Gene ID: 266766) in rat. Calcipressin-1 is a cytoplasmic protein that directly binds to the catalytic domain of calcineurin and acts as its negative regulator. When phosphorylated at the conserved FLISPP motif its capacity to inhibit calcineurin is increased. This phosphorylation also controls the half-life of calcipressin-1 by accelerating its degradation. It is a calcium-responsive enzyme, which is reported to dephosphorylate the nuclear factor of activated T cells (NFAT), which promotes its nuclear translocation and links calcium signaling to transcriptional regulation. Calcipressin-1 displays a quick turnover with a half-life of less than 3 hours. However, a Ser to Ala mutation at Ser163 prolongs its half-life to more than 9 hours. (Ref.: Genesca, L et al. (2003). Biochem. J. 374(2); 567-575).

Specificity

This rabbit polyclonal antibody detects calcipressin-1 (RCAN-1) in human and rat cells. It targets an epitope with in 11 amino acids from the C-terminal half.

Immunogen

A linear peptide corresponding to 11 amino acids from the C-terminal half of rat Calcipressin-1.
Epitope: unknown

Application

Anti-Calcipressin-1 Antibody, Cat. No. ABN1676, is a highly specific rabbit polyclonal antibody that targets Calcipressin-1 and has been tested in Immunocytochemistry and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A representative lot detected Calcipressin-1 in cultured rat sympathetic neurons (Patel, A., et. al. (2015). Nat Commun. 6:10119).

Immunocytochemistry Analysis: A representative lot detected Calcipressin-1 in sympathetic neurons (Patel, A., et. al. (2015). Nat Commun. 6:10119).

Quality

Evaluated by Immunocytochemistry in SH-SY5Y cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Calcipressin-1 in SH-SY5Y cells.

Target description

22.95 kDa calculated.

Physical form

Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service