Skip to Content
Merck
All Photos(2)

Key Documents

ABN1018

Sigma-Aldrich

Anti-LRP1 Antibody, Intracellular domain

serum, from rabbit

Synonym(s):

Prolow-density lipoprotein receptor-related protein 1, A2MR, Alpha-2-macroglobulin receptor, APOER, Apolipoprotein E receptor, CD91, LRP-1, LRP-85, LRPICD

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 10,200.00

CZK 10,200.00


Estimated to ship onApril 04, 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1311

Request a Bulk Order

Select a Size

Change View
100 μL
CZK 10,200.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 10,200.00


Estimated to ship onApril 04, 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1311

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... LRP1(4035)

General description

Prolow-density lipoprotein receptor-related protein 1 (UniProt Q07954; also known as A2MR, Alpha-2-macroglobulin receptor, APOER, Apolipoprotein E receptor, CD91, LRP-1) is encoded by the LRP1 (also known as A2MR, APR) gene (Gene ID 4035) in human. LRP1 is an LDLR family member expressed by vascular endothelial cells forming the blood-brain barrier (BBB), vascular mural cells (pericytes and vascular smooth muscle cells (VSMCs)), neurons, and astrocytes. LRP1 recognizes and mediates the endocytosis of more than 40 different ligands, including apolipoprotein E (apoE), APP and Aβ. LRP1 is the primary receptor mediating transport of Aβ across the BBB into circulation, thereby clearing it from the brain. LRP1 is synthesized with a signal peptide sequence (a.a. 1-19), posttranslational cleavage yields a soluble form (sLRP1) and a cell surface-bound form composed of an 515 kDa extracellular α-chain (LRP-515) and a 85 kDa single-transmembrane (a.a. 4420-4444) β-chain (LRP-85) that contains the 100-amino intracellular domain (LRPICD; a.a. 4445-4544). BACE-1, ADAM-10, ADAM-12, ADAM-17, MT1-MMP, and t-PA are shown to be involved in the shedding of LRP1 to produce sLRP1.

Specificity

Recognizes the Pro- and mature LRP-1 (a.a. 1-4544 and 20-4544, respectively), the beta-secretase-cleaved, membrane-bound LRP-85, and the gamma-secretase-cleaved intracellular domain LRPICD fragment, but not the beta-secretase-cleaved extracellular LRP-515 fragment.

Immunogen

Epitope: LRPICD.
GST-tagged recombinant human LRP1 intracellular domain (LRPICD).

Application

Anti-LRP1 Antibody, Intracellular domain is an antibody against LRP1 for use in Western Blotting.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Western Blotting Analysis: A representative lot detected the LRP-85 target band in NIH/3T3 cell lysate, as well as in mouse brain and liver homogenates (Courtesy of Professor Nicola Zambrano, University of Naples Federico II, Italy).
Western Blotting Analysis: A representative lot detected a decreased LRP-85 target band in lysates from LRP1 shRNA-treated NIH/3T3 cells (Courtesy of Professor Nicola Zambrano, University of Naples Federico II, Italy).

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected the LRP-85 target band in 10 µg of NIH/3T3 cell lysate.ysate.

Target description

~80 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Rabbit polyclonal antibody serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service