Skip to Content
Merck
All Photos(2)

Key Documents

ABE491

Sigma-Aldrich

Anti-Histone-lysine N-methyltransferase NSD3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone-lysine N-methyltransferase NSD3, Nuclear SET domain-containing protein 3, Protein whistle, WHSC1-like 1 isoform 9 with methyltransferase activity to lysine, Wolf-Hirschhorn syndrome candidate 1-like protein 1, WHSC1-like protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
CZK 10,800.00

CZK 10,800.00


Estimated to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
200 μG
CZK 10,800.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 10,800.00


Estimated to ship onApril 07, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WHSC1L1(54904)

General description

Histone-lysine N-methyltransferase NSD3, also known as Nuclear SET domain-containing protein 3, Protein whistle, WHSC1-like 1 isoform 9 with methyltransferase activity to lysine or Wolf-Hirschhorn syndrome candidate 1-like protein 1, (WHSC1-like protein 1), and encoded by the gene WHSC1L1/NSD3, is a histone methylase that preferentially methylates Lys-4 and Lys-27 of histone H3. Lys-4 methylation is generally a specific tag for transcriptional activation while Lys-27 methylation is a mark for transcriptional repression. NSD3 is highly expressed in many tissues including the brain, heart and skeletal muscle. Overexpression of NSD3 is associated with certain forms of lung cancer as well as certain other cancers. NSD3 also facilitates the expression of many other genes important in cancer progression such as Brd4. Brd4 through its extraterminal domain interacts with NSD3 which regulates transcription by modifying the chromatin microenvironment at Brd4 target genes.

Specificity

This antibody is expected to cross react with isofomrs 1, 2, and 4 based upon immunogen design.

Immunogen

KLH-conjugated linear peptide corresponding to a region near the N-terminus of human Histone-lys N-methyltransferase NSD3.

Application

Detect Histone-lysine N-methyltransferase NSD3 using this rabbit polyclonal antibody, Anti-Histone-lysine N-methyltransferase NSD3 Antibody validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected Histone-lys N-methyltransferase NSD3 in human and mouse cerebellum tissues.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blotting in BALB/3T3 nuclear extract.

Western Blotting Analysis: 2 µg/mL of this antibody detected Histone-lys N-methyltransferase NSD3 in 10 µg of BALB/3T3 nuclear extract.

Target description

~180 kDa, ~160 kDa, ~90 kDa, and ~70 kDa observed. Uniprot desribes four isoforms at 162 kDa (isoform 1), 156 kDa (isoform 2), 73 kDa (isoform 3), and isoform 4 (155 kDa). This protein is known to be acetylated and phosphorylated and may be observed at higher molecular weights. Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service