Skip to Content
Merck

Skip To

ABE335

Anti-FOXL2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Forkhead box protein L2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship on21 August 2026fromAreál Kühne+Nagel spol. s r.o.

11 500,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-

11 500,00 CZK


Estimated to ship on21 August 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), rabbit (based on 100% sequence homology), mouse (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Forkhead box protein L2 (FOXL2) is a fork-head transcription factor that is expressed in developing gonad and in granulosa cells of the mature ovary. It plays a role in repressing the development of testis, and facilitating the differentiation and maintenance of the ovary. Mutations in the FOXL2 gene result in blepharophimosis ptosis epicanthus inversus (BPES) syndrome, which is characterized by craniofacial defects and premature ovarian failure. FOXL2 has also been implicated in the development of ovarian granulosa cell tumors.
~50 kDa observed. An uncharacterized band may appear at ~25 kDa in some lysates. The calculated molecular weight is 39 kDa, however FOXL2 has been shown as a ~50 kDa band in western blots (Marongiu, M., et al. (2010). PLOS One. 5(3):1-9).

Immunogen

KLH-conjugated linear peptide corresponding to human FOXL2.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This Anti-FOXL2 Antibody is validated for use in Western Blotting for the detection of FOXL2.

Biochem/physiol Actions

Other homologies: Chicken (77% sequence homology).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human pituitary
Evaluated by Western Blot in human pituitary tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected FOXL2 in 10 µg of human pituitary tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AV39509AV31709AV31694
description

from rabbit, purified by affinity chromatography

description

IgG fraction of antiserum

description

IgG fraction of antiserum

description

IgG fraction of antiserum

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

IgG fraction of antiserum

antibody form

IgG fraction of antiserum

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

technique(s)

western blot: suitable

technique(s)

western blot: suitable

technique(s)

western blot: suitable

technique(s)

immunohistochemistry: suitable, western blot: suitable


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters