Skip to Content
Merck

Skip To

ABE2863M

Anti-E3 ubiquitin-protein ligase (UBR5/EDD)

serum, from rabbit

Synonym(s):

E3 ubiquitin-protein ligase UBR5, EDD, EC:2.3.2.26, E3 ubiquitin-protein ligase, HECT domain-containing 1, HECT-type E3 ubiquitin transferase UBR5, Hyperplastic discs protein homolog, hHYD, Progestin-induced protein, EDD

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL

Estimated to ship on20 November 2026fromAreál Kühne+Nagel spol. s r.o.

10 100,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IP, WB
Citations:
-

10 100,00 CZK


Estimated to ship on20 November 2026Details


​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

technique(s)

immunofluorescence: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... UBR5(51366)

General description

E3 ubiquitin-protein ligase UBR5 (UniProt: O95071; also known as EC:2.3.2.26, E3 ubiquitin-protein ligase, HECT domain-containing 1, HECT-type E3 ubiquitin transferase UBR5, Hyperplastic discs protein homolog, hHYD, Progestin-induced protein) is encoded by the UBR5 (also known as EDD, EDD1, HYD, KIAA0896) gene (Gene ID: 51366) in human. UBR5 is a nuclear protein that is widely expressed and is shown to be most abundant in testis and is expressed at high levels in brain, pituitary, and kidney. Two isoforms of UBR5 are described that are produced by alternative splicing and differ only in a single amino acid. It is identified as a novel regulator of p53 and downregulation of UBR5 results in elevated levels of p53 in normal and transformed cells. Its deficiency in mice is reported to result in mid gestation embryonic death due to a compromised yolk sac vascular development. UBR5 is a component of the N-end rule pathway. It recognizes and binds to proteins bearing specific N-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation. It is shown to be involved in maturation and/or transcriptional regulation of mRNA by activating CDK9 by polyubiquitination and is believed to play a role in control of cell cycle progression. It associates with CDK9 and TFIIS/TCEA1 and forms a transcription regulatory complex made of CDK9, RNAP II, UBR5, and TFIIS/TCEA1 that can stimulate target gene transcription. UBR5 regulates DNA topoisomerase II binding protein (TopBP1) in the DNA damage response and may have a role in tumor suppression. (Ref.: Smits, VAJ (2012). Cell Cycle 11(4); 715-720).
~309 kDa observed; 309.35 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: unknown
His-tagged recombinant fragment corresponding to 331 amino acids from the internal region of human E3 ubiquitin-protein ligase UBR5/EDD.

Application

Anti-E3 ubiquitin-protein ligase (UBR5/EDD), Cat. No. ABE2863, is a highly specific rabbit polyclonal antibody that targets E3 ubiquitin-protein ligase UBR5 and has been tested in Immunofluorescence, Immunoprecipitation, and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A 1:50,000 dilution from a representative lot detected E3 ubiquitin-protein ligase (UBR5/EDD) in 10 µg of Balb 3T3/A31 cell lysate.

Western Blotting Analysis: A representative lot detected E3 ubiquitin-protein ligase (UBR5) in U2OS cells transefected with Luc or different EDD siRNA oligos (Smits, V.A., et. al. (2012). Cell Cycle. 11(4):715-20).

Immunoprecipitation Analysis: 2.5ul µL from a representative lot detected E3 ubiquitin-protein ligase (UBR5?EDD) in U2OS cell lysate (Courtesy of Dr. Veronique Smits at Hospital Univers. Canarias, Spain).

Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected E3 ubiquitin-protein ligase (UBR5/EDD) in U2OS cells (Courtesy of Dr. Veronique Smits at Hospital Univers. Canarias, Spain).

Biochem/physiol Actions

This rabbit polyclonal antibody detects E3 ubiquitin-protein ligase UBR5/EDD in human and murine cells. It targets an epitope within 331 amino acids from the internal region.

Physical form

Rabbit polyclonal antiserum with 0.1% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in U2OS cell lysate.

Western Blotting Analysis: A 1:50,000 dilution of this antibody detected E3 ubiquitin-protein ligase (UBR5/EDD) in 10 µg of U2OS cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABF1110ZRB1698HPA053688
description

serum, from rabbit

description

clone 16G10.1, from mouse

description

recombinant, expressed in HEK 293 cells

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

serum

antibody form

purified immunoglobulin

antibody form

purified antibody

antibody form

affinity isolated antibody

biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

UniProt accession no.

O95071

UniProt accession no.

O95071

UniProt accession no.

O95071

UniProt accession no.

O95071

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable, immunofluorescence: suitable

technique(s)

western blot: suitable, immunohistochemistry: suitable (paraffin)

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, immunocytochemistry: suitable, affinity binding assay: suitable, flow cytometry: suitable, western blot: suitable

technique(s)

immunofluorescence: 0.25-2 μg/mL


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters