Skip to Content
Merck

ABE2580

Anti-Histone H3Kme3Q5Ser

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
mouse, human
Application:
ChIP, DB, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibody form

purified antibody

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

ChIP: suitable, dot blot: suitable, western blot: suitable

isotype

IgG

UniProt accession no.

General description

Histone H3.1 UniProt: Q16695; also known as H3/t, H3t, H3/g) is encoded by H3-4 (also known as H3FT, HIST3H3) gene (Gene ID: 8290) in human. Histone H3 has two main variants, H3.1 and H3.3, which show different genomic localization patterns in animals. Histone H 3.1 is a core component of nucleosome that is present only in mammals. It is expressed during S phase, then its expression decreases significantly as cell division slows down during the process of differentiation. Histone H 3.1 expression is shown to be replication dependent. It serves as the canonical histone, which is incorporated during DNA replication. Histone 3.1 can also undergo serotonylation of glutamine at position 5 (Q5ser) in organisms that produce serotonin. This has been demonstrated that tissue transglutaminase 2 can serotonylate histone H3K4me3 marked nucleosomes, which results in the formation of H3K4me3Q5ser. The presence of Q5ser does not affect the ability of MLL1, an H3K4 methyltransferase, to modify nucleosomes in vitro. In the brain the H3K4me3Q5ser signal is not segregated only to regions in which serotonin is produced, but is shown to be broadly distributed. H3K4me3Q5ser expression has also been reported in human pluripotent stem cell-derived serotonergic neurons and with differentiation the levels of H3K4me3Q5ser increase significantly. This polyclonal Anti-Histone H3Kme3Q5Ser is suitable for Western Blotting, Dot blotting, ChIP seq applications. (Ref.: Farrelly, L.A., et al. (2019). Nature. 567(7749):535-539).
Target molecular weight ~16 kDa observed; 15.51 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids from the N-terminal region of human Histone H3 trimethylated at lysine 4 and serotonylated at glutamine 5.

Application

Epitope: N-terminus
Tested Applications

Western Blotting Analysis: A 1:500 dilution from a representative lot detected Histone H3 trimethylated at Lys4 and serotonylated at Gln5 in whole cell lysate from differentiated RN46A-B14 cells but not in lysate from undifferentiated RN46A-B14 cells.

ChIP-Seq Analysis: A representative lot of this antibody was used to assess genome-wide impacts of serotonergic differentiation in differentiated RN46A-B14 cells (Farrelly, LA., et al. (2109). Nature. 567(7749); 535-539).

Western Blotting Analysis; A representative lot of this antibody was used to detect Histone H3 trimethylated at Lys4 and serotonylated at Gln5 by Western Blotting application. (Farrelly, LA., et al. (2109). Nature. 567(7749); 535-539).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Anti-Histone H3Kme3Q5Ser, Cat. No. ABE2580, is a rabbit polyclonal antibody that detects Histone H3Kme3Q5Ser and is tested for use in Dot Blot and ChIP-seq, and Western Blotting.

Physical form

Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Recommended storage: +2°C to +8°C.

Analysis Note

Evaluated by Dot Blot with unmodified and modified Histone H3Kme3Q5Serotonyl peptides.

Dot Blot Analysis: A 1:500 dilution from a representative lot detected Histone H3 trimethylated at Lys4 and serotonylated at Gln5 but not Histone H3(1-10) control synthetic peptide.

Other Notes

Human, Mouse.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library