Skip to Content
Merck

Skip To

ABE1975

Anti-Srg3/BAF155 Antibody

serum, from rabbit

Synonym(s):

SWI/SNF complex subunit SMARCC1, BAF155, BRG1-associated factor 155, SWI/SNF complex 155 kDa subunit, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily C member 1, SWI3-related protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL

Estimated to ship onJune 05, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 10,500.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ChIP, WB
Citations:
-

CZK 10,500.00


Estimated to ship onJune 05, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

ChIP: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SMARCC1(6599)

General description

SWI/SNF complex subunit SMARCC1 (UniProt P97496; also known as BAF155, BRG1-associated factor 155, SWI/SNF complex 155 kDa subunit, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily C member 1, SWI3-related protein) is encoded by the Smarcc1 (also known as Baf155, Srg3) gene (Gene ID 20588) in murine species. SMARCC1/BAF155/SRG3 is a core component of the SWI/SNF complex that coordinates the expression of specific genes crucial for germinal center (GC) formation, including Blimp-1 and Bcl-6. The SWI/SNF is a chromatin-remodeling complex that drives conformational changes of nucleosomes in an ATP-dependent manner. SWI/SNF complex is required for the differentiation of GC B and T follicular helper (Tfh) cells. Smarcc1/Baf155/Srg3-knockout mice showed impaired differentiation of GC B and Tth cells in response to T cell-dependent antigen challenge. The SWI/SNF complex consists of at least 12 different subunits, including BRG1 with ATPase activity and SMARCC1/BAF155/SRG3 that controls the complex stability. SMARCC1/BAF155/SRG3 contains an N-terminal acidic region (23% Asp/Glu content from a.a. 211-441), a DNA-binding SWIRM domain (a.a. 448-545), a histone-interacting SANT domain (a.a. 617-668), a leucine-zipper motif (a.a. 899-942), and a C-terminal glutamine- and proline-rich region (a.a. 768-862 & 977-1104).
~155 kDa observed. 122.9 kDa (human and mouse SRG3) and 120.1 kDa (mouse spliced isoform 2) calculated. Uncharacterized band(s) may appear in some lysates.

Immunogen

Epitope: C-terminal region.
GST-tagged recombinant mouse Srg3/BAF155 C-terminal fragment.

Application

Anti-Srg3/BAF155 Antibody is an antibody against Srg3/BAF155 for use in Western Blotting, Chromatin Immunoprecipitation (ChIP).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Western Blotting Analysis: A representative lot detected Srg3 immunoprecipitated from the nuclear extracts of human Raji and murine BAL17 B cells (Choi, J., et al. (2015). Proc. Natl. Acad. Sci. U. S. A. 112(7):E718-727).
Western Blotting Analysis: A representative lot detected the ~155 kDa target Srg3 band and a ~170 kDa cross-reactive band in murine thymus and lymph node lysates. The ~170 kDa band likely represents the murine ortholog of human BAF170 protein (Jeon, S.H., et al. (1997). J. Exp. Med. 185(10):1827-1836).
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected Srg3 occupancy at the proximal promoter and BRE (Bcl-6 responsive element) regions of the Blimp-1 locus in germinal center (GC) B cells, but not in plasma B cells (PCs) (Choi, J., et al. (2015). Proc. Natl. Acad. Sci. U. S. A. 112(7):E718-727).
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected Srg3 occupacy at the Blimp-1 locus using chromatin preprations from untreated murine BAL17 B lymphoma cells, but not Brg1/Srg3-depleted BAL17 cells following siRNAs treatment (Choi, J., et al. (2015). Proc. Natl. Acad. Sci. U. S. A. 112(7):E718-727).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Srg3 and its associated proteins from the nuclear extracts of human Raji and murine BAL17 B cells (Choi, J., et al. (2015). Proc. Natl. Acad. Sci. U. S. A. 112(7):E718-727).
Immunoprecipitation Analysis: A representative lot co-immunoprecipitated a ~195 kDa SWI2-like protein with the ~155 kDa Srg3 protein from murine thymus cell extracts (Jeon, S.H., et al. (1997). J. Exp. Med. 185(10):1827-1836).

Biochem/physiol Actions

Reacts with both mouse and human Srg3/BAF155 proteins. Reproted to exhibit cross-reactivity toward a ~170 kDa murine ortholog of human BAF170 protein (Jeon, S.H., et al. (1997). J. Exp. Med. 185(10):1827-1836). Expected to react with also murine Srg3/Smarcc1 spliced isoform 2 reported by UniProt (UniProt P97496-2).

Physical form

Rabbit polyclonal antibody serum with 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected Srg3/BAF155 in 10 µg of K562 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA026853MABE121WH0006598M1
antibody form

serum

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

mouse

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

species reactivity

human, mouse

species reactivity

human

species reactivity

rat, human

species reactivity

mouse

shipped in

dry ice

shipped in

wet ice

shipped in

wet ice

shipped in

dry ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters