Skip to Content
Merck

Skip To

ABE1877M

Anti-PAXIP1

from rabbit

Synonym(s):

PAX-interacting protein 1, PAX transactivation activation domain-interacting protein, PAXIP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL

Estimated to ship onJune 03, 2027fromAreál Kühne+Nagel spol. s r.o.

CZK 9,780.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ChIP, IP, WB
Citations:
-

CZK 9,780.00


Estimated to ship onJune 03, 2027Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

technique(s)

ChIP: suitable (ChIP-seq), immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PAXIP1(22976)
mouse ... Paxip1(55982)

General description

PAX-interacting protein 1 (UniProt: Q6NZQ4; also known as PAX transactivation activation domain-interacting protein, PAXIP1) is encoded by the Paxip1 (also known as Ptip) gene (Gene ID: 55982) in murine species. PAXIP1 is a nuclear matrix protein that is involved in DNA damage response and in transcriptional regulation through histone methyltransferase (HMT) complexes such as the MLL2/MLL3 complex. In vitro it is shown to be involved in the homologous recombination mechanism for the repair of double-strand breaks. It recruits Tp53bp1 to DNA damage foci and effective DNA damage response appears to require the association with Tp53bp1 phosphorylated by ATM. It also recruits PAGR1A (Pax-Interacting Protein 1-associated glutamate rich protein 1a) to DNA damage sites and PAGR1A:PAXIP1 complex is believed to be essential for cell survival. PAXIP1 expression is reported in all tissues and high expression is observed in embryonic kidney and brain. PAXIP1 contains 6 BRCT domains and BRCT domains 1 and 2 mediate its interaction with PAGR1A and interaction with MLL2/MLL3 is accomplished with BRCT domains 5 and 6. Mice with defective Paxip1 gene are developmentally retarded and usually face embryonic lethality by day 9.5 (E9.5).
~51 kDa observed (recombinant); 119.27 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: unknown
His-tagged recombinant fragment corresponding to 199 amino acids from the N-terminal region of murine PAX-interacting protein 1.

Application

Anti-PAXIP1, Cat. No. ABE1877, is a highly specific rabbit polyclonal antibody that targets PAX-interacting protein 1 and has been tested for use in ChIP-seq and Chromatin Immunoprecipitation (ChIP), Immunoprecipitation, and Western Blotting.
Chromatin Immunoprecipitation Analysis: A representative lot detected PAXIP1 in mouse embryonic fibroblasts (Cho, Y.W., et. al. (2009). Cell Metab. 10(1):27-39).

ChIP-seq Analysis: Analysis: A representative lot detected PAXIP1 in ChIP-seq applications (Callen, E., et. al. (2012). Immunity. 37(6):971-85).

Immunoprecipitation Analysis: A representative lot detected PAXIP1 in Immunoprecipitation applications (Gong, Z., et. al. (2009). J Biol Chem. 284(11):7284-93).

Immunoprecipitation Analysis: 1 µg from a representative lot detected PAXIP1 in nuclear extract of wild type and PTIP KO mouse embryonic fibroblasts (Courtesy of Dr. Ji-Eun Lee and Dr. Kai Ge at NIH).

Western Blotting Analysis: A representative lot detected PAXIP1 in Western Blotting applications (Cho, Y.W., et. al. (2009). Cell Metab. 10(1):27-39; Gong, Z., et. al. (2009). J Biol Chem. 284(11):7284-93).
Research Category
Epigenetics & Nuclear Function

Biochem/physiol Actions

This rabbit polyclonal antibody detects PAX-interacting protein 1 in human and murine cells. It targets an epitope within 199 amino acids from the N-terminal region.

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting with with recombinant Pax-interacting protein 1.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected recombinant PAX-interacting protein 1.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABE69HPA006694ABE1863
biological source

rabbit

biological source

chicken

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

species reactivity

human, mouse

species reactivity

rat, mouse, human

species reactivity

human

species reactivity

mouse, human

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters