Skip to Content
Merck

Skip To

ABE1860

Anti-CDK13

from rabbit

Synonym(s):

Cyclin-dependent kinase 13, CDC2-related protein kinase 5, Cell division cycle 2-like protein kinase 5, Cell division protein kinase 13, hCDK13, Cholinesterase-related cell division controller

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
2
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CDK13(8621)

General description

Cyclin-dependent kinase 13 (UniProt: Q14004: also known as Cdk13, EC:2.7.11.22, EC:2.7.11.23, CDC2-related protein kinase 5, Cell division cycle 2-like protein kinase 5, Cell division protein kinase 13, hCDK13, Cholinesterase-related cell division controller) is encoded by the CDK13 (also known as CDC2L, CDC2L5, CHED, KIAA1791) gene (Gene ID:8621) in human. CDK13 is a member of the cyclin-dependent serine/threonine protein kinase family. Members of this family are known to play a vital role as master switches in cell cycle control. Human CDK 13 shares 43% sequence identity with CDK12. CDK13 is expressed in fetal brain, liver, muscle, in adult brain and in neuroblastoma and glioblastoma tumors. It contains expanded regions of serine-arginine (SR) motifs in its N-terminal regions, which links it to the SR protein family involved in RNA processing and pre-mRNA splicing. CDK13 contains a C-terminal extension helix that is composed of a polybasic cluster and a DCHEL motif that interacts with the bound ATP. CDK13 displays C-terminal repeat domain (CTD) kinase activity and is required for RNA splicing. It hyperphosphorylates the C-terminal heptapeptide repeat domain of the largest RNA polymerase II subunit RPB1 and acts a key regulator of transcription elongation. Phosphorylation of the three serine residues (Ser 2, 5, and 7) within the CTD is linked to the phases of RNAPII-mediated transcription. Phosphorylation of Ser7 (pSer7) starts the transcription cycle that primes the CTD for further modifications. (Ref.:Greifenberg, AK et al. (2016). Cell Reports 14(2): 320-331).
~170 kDa observed; 164.92 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminal half of human Cyclin-dependent kinase 13 (CDk13).

Application

Anti-CDK13, Cat. No. ABE1860, is a highly specific rabbit polyclonal antibody that targets Cyclin-dependent kinase 13 and has been tested in Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A representative lot detected CDK13 in HeLa cell lysate (Bartkowiak, B., et. al. (2010). Genes Dev. 24(20):2303-16).

Western Blotting Analysis: 1:1,000 dilution from a representative lot detected CDK13 in HEK293 whole cell extract. (Courtesy of Dr. A. Greenleaf, Duke University Medical Center).

Western Blotting Analysis: 1:1,000 dilution from a representative lot detected CDK13 in HeLa nuclear extract. (Courtesy of Dr. A. Greenleaf, Duke University Medical Center).

Western Blotting Analysis: 1:1,000 dilution from a representative lot detected CDK13 in OVCA5 and HEK293 (Courtesy of Dr. A. Greenleaf, Duke University Medical Center).


Biochem/physiol Actions

This polyclonal antibody specifically detects CDK13 in HEK293 cells.

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HEK293 whole cell extract.

Western Blotting Analysis: 1:500 dilution of this antibody detected CDK13 in 20 µL of HEK293 whole cell extract.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters