Skip to Content
Merck

ABE125

Anti-PRMT7 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protein arginine N-methyltransferase 7, Histone-arginine N-methyltransferase PRMT7, Myelin basic protein]-arginine N-methyltransferase PRMT7

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

feline (based on 100% sequence homology), bovine (based on 100% sequence homology), ferret (based on 100% sequence homology), horse (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

General description

PRMT7, also known as Protein arginine N-methyltransferase 7, Histone-arginine N-methyltransferase PRMT7 or Myelin basic protein-arginine N-methyltransferase PRMT7, and encoded by the gene PRMT7/KIAA1933, is an arginine methyltransferase that can both catalyze the formation of omega-N monomethylarginine (MMA) and symmetrical dimethylarginine (sDMA), with a preference for the formation of MMA. PRMT7 specifically mediates the symmetrical dimethylation of arginine residues in the small nuclear ribonucleoproteins SmD1 (SNRPD1) and SmD3 (SNRPD3). Such methylation is required for the assembly and biogenesis of snRNP core particles. PRMT7 also mediates the symmetric dimethylation of histone H4 ′Arg-3′ to form H4R3me2s, and it plays a role in gene imprinting by being recruited by CTCFL at the H19 imprinted control region (ICR) and methylating histone H4 to form H4R3me2s and thus leading to the recruitment of DNA methyltransferases to those sites. Finally, PRMT7 may also play a role in embryonic stem cell (ESC) pluripotency and is able to methylate histone H2A and myelin basic protein (MBP).
~75 kDa observed. Uniprot describes four isoforms at 79 kDa (Isoform 1), 62 kDa (Isoform 2), 73 kDa (Isoform 3), and 64 kDa (Isoform 4).

Immunogen

Epitope: unknown
GST-tagged recombinant protein corresponding to human PRMT7

Application

Anti-PRMT7 Antibody detects level of PRMT7 & has been published & validated for use in PRMT7.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Biochem/physiol Actions

This antibody is expected to cross react with all isoforms based upon immunogen design.

Physical form

Affinity purified
Purified rabbit Polyclonal serum in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in U2OS nuclear extract.

Western Blotting Analysis: A 1:500 dilution of this antibody detected PRMT7 in 10 µg of U2OS nuclear extract.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10-13 - German Storage Class 10 to 13



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library