Skip to Content
Merck

Skip To

ABE1044

Anti-HSF 1 Antibody

serum, from rabbit

Synonym(s):

Heat shock factor protein 1, HSF 1, Heat shock transcription factor 1, HSTF 1

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ChIP, IP, WB
Citations:
12


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

ChIP: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HSF1(3297)

General description

HSF1, also known as Heat shock factor protein 1 (HSF-1) or Heat shock transcription factor 1 (HSTF-1), and encoded by the gene HSF1/HSTF1, is a DNA binding transcription factor that binds heat shock promoter elements (HSE’s) and activates transcription. In higher eukaryotes, HSF1 is unable to bind to HSE regions unless the cells are heat shocked. HSF1 is normally a monomer that interacts with HSP90 and other chaperones in a large complex in the cytoplasm. Upon heat shock HSF1 is released from the complex, dimerizes, and is transported into the nucleus. HSF1 is localized to the cytoplasm during normal growth the protein localizes to the nucleus upon heat shock. HSF1 is part of the cellular stress response, and HSF1 is known to interact with many other stress- induced proteins including NCOA6, SYMPK, RALBP1, HSF2, CEBPB, HSPA1A, Heat shock protein 90kDa alpha and HSPA.
~75 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Immunogen

Recombinant protein corresponding to mouse HSF 1.

Application

Anti-HSF 1 antibody is an antibody against HSF 1 for use in western blotting, IP & ChIP.
Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated HSF 1 from HEK293 cell lysates transfected with wild type or mutant hRP1-HA (Fujimoto, M., et al. (2012). Mol Cell. 48(2):182-194.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated HSF 1 from hHSF1 transfected MEF cell lysate (Fujimoto, M., et al. (2010). Mol Cell Biol. 21(1):106-116.).

Chromatin Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated HSF 1 from wild type and HSF2-null primary MEF cell lysates (Shinkawa, T., et al. (2011). Mol Cell Biol. 22(19):3571-3583.).

Chromatin Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated HSF 1 from HeLa and MEF cell lysates which were either untreated or heat shock treated (Prof. A. Nakai, Yamaguchi University School of Medicine).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting: A 1:1,000 dilution of this antibody detected HSF 1 in 10 µg of HeLa cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters