Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μL | Estimated to ship onJune 05, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 10,900.00 |
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
technique(s)
immunohistochemistry: suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... UHRF2(115426)
General description
Immunogen
Application
Epigenetics & Nuclear Function
Nuclear Receptors
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: A 1:5,000 dilution of this antibody detected E3 ubiquitin-protein ligase UHRF2 in 10 µg of WI-38 cell lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| shipped in dry ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| Gene Information human ... UHRF2(115426) | Gene Information human ... UHRF2(115426) | Gene Information human ... UHRF2(115426) | Gene Information human ... UHRF2(115426) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



