Skip to Content
Merck

Skip To

ABC834

Anti-ZIP-6 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Zinc transporter ZIP6, Estrogen-regulated protein LIV-1, Solute carrier family 39 member 6, Zrt- and Irt-like protein 6, ZIP-6

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship on10 July 2026fromAreál Kühne+Nagel spol. s r.o.

11 800,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-

11 800,00 CZK


Estimated to ship on10 July 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

technique(s)

immunohistochemistry: suitable, western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLC39A6(25800)

General description

Zinc transporter ZIP6, also known as Estrogen-regulated protein LIV-1 or Solute carrier family 39 member 6 orZrt- and Irt-like protein 6 (ZIP-6), and encoded by the gene name ZIP6 or SLC39A6 or LIV1 is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP-6 was initially identified as LIV-1, an estrogen-regulated gene that has been implicated in metastatic breast cancer. Elevated ZIP-6 expression has also been reported in human cervical cancer and the HeLa cell line; down-regulation of ZIP-6 expression in HeLa by RNAi inhibited cell proliferation, colony formation, migration and invasiveness, as well as decreasing Snail and Slug levels, suggesting ZIP-6 plays a regulatory role on the ERK1/2-Snail/Slug pathway.
~48 and ~85 kDa observed. Uncharacterized bands may be observed in some lysate(s). Uniprot describes 2 isoforms produced by alternative splicing at ~49 kDa and ~85 kDa

Immunogen

Epitope: center region
KLH-conjugated synthetic peptide corresponding to the center region of human Anti-ZIP-6.

Application

Anti-ZIP-6 Antibody detects level of ZIP-6 & has been published & validated for use in ZIP-6.
Immunohistochemistry Analysis: 5 μg/mL from a representative lot detected Anti-ZIP-6 in mouse lung tissue.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Biochem/physiol Actions

This antibody is recognizes isoforms at ~48 and ~85 kDa and is predicted to not cross-react with other ZIP family members.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in mouse lung tissue lysate.

Western Blotting Analysis: 1 μg/mL of this antibody detected Anti-ZIP-6 in mouse lung tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA042377ABF197AV44019
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Gene Information

human ... SLC39A6(25800)

Gene Information

human ... SLC39A6(25800)

Gene Information

human ... SLC39A8(64116)

Gene Information

human ... SLC30A1(7779)

UniProt accession no.

Q13433

UniProt accession no.

Q13433

UniProt accession no.

Q9C0K1

UniProt accession no.

Q9Y6M5


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters