Skip to Content
Merck

Skip To

ABC1688

Anti-SWAP-70

from rabbit, purified by affinity chromatography

Synonym(s):

Switch-associated protein 70

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 μg

Estimated to ship on03 August 2026fromAreál Kühne+Nagel spol. s r.o.

4 040,00 CZK
100 μg

Estimated to ship on03 August 2026fromAreál Kühne+Nagel spol. s r.o.

11 200,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-

4 040,00 CZK


Estimated to ship on03 August 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), bovine (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... SWAP70(23075)

General description

Switch-associated protein 70 (UniProt: Q9UH65; also known as SWAP-70) is encoded by the SWAP70 (also known as KIAA0640, HSPC321) gene (Gene ID: 23075) in human. SWAP-70 is a phosphatidylinositol 3,4,5-trisphosphate (PIP3)-dependent guanine nucleotide exchange factor (GEF) that, independently of RAS, transduces signals from tyrosine kinase receptors to RAC. SWAP-70 regulates the actin cytoskeleton as an effector or adapter protein in response to agonist stimulated phosphatidylinositol (3,4)-bisphosphate production and cell protrusion. It is expressed only in mature B-cells including those associated with mucosa-associated tissue and bronchus-associated tissue. In resting B-cells it is localized mainly in the cytoplasm and upon cell activation it is recruited to the plasma membrane and then translocates to the nucleus. In activated, class-switching B-cells it is associated with membrane IgG, but not IgM. It is also widely expressed in spleen, kidney, lung, and liver. SWAP-70 has an N-terminal putative EF-hand domain, a pleckstrin homology (PH) domain (aa 210-306) that binds phosphatidylinositol 3,4,5-trisphosphate and is responsible for membrane localization, a tri-partite coiled-coil region (aa 316-539), and a C-terminal F-actin-binding site through which it specifically binds non-muscle actin. (Ref.: Chacón-Martínez, CA et al. (2103). J. Biol. Chem. 288(40); 28687-28703).
~70 kDa observed; 69.00 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

KLH-conjugated linear peptide corresponding to 12 amino acids from the N-terminal region of human Switch-associated protein 70.

Application

Anti-SWAP-70, Cat. No. ABC1688, is a rabbit polyclonal antibody that detects Switch-associated protein 70 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected SWAP-70 in human tonsil and human lung tissue sections.

Biochem/physiol Actions

This rabbit polyclonal antibody detects human Switch-associated protein 70. It targets an epitope with in 12 amino acids from the N-terminal region.

Analysis Note

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected SWAP-70 in Raji cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4503107MABF920ZRB1717
description

from rabbit, purified by affinity chromatography

description

affinity isolated antibody

description

clone 1A9, from rat

description

recombinant, expressed in HEK 293 cells

biological source

rabbit

biological source

rabbit

biological source

rat

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified antibody

antibody form

purified antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Gene Information

human ... SWAP70(23075)

Gene Information

human ... ZAP70(7535)

Gene Information

human ... SH2D1A(4068)

Gene Information

human ... ZAP70(7535)

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

technique(s)

ELISA: 1:1000, immunohistochemistry: 1:50-1:100

technique(s)

flow cytometry: suitable, western blot: suitable

technique(s)

flow cytometry: suitable, immunocytochemistry: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, peptide synthesis: suitable, western blot: suitable

UniProt accession no.

Q9UH65

UniProt accession no.

P43403

UniProt accession no.

O88890

UniProt accession no.

P43403


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters