Skip to Content
Merck

ABC149

Anti-BST2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Bone marrow stromal antigen 2, BST-2, HM1.24 antigen, Tetherin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BST2(684)

General description

BST-2 also known as CD317 and Tetherin, bone marrow stromal antigen 2 (BST-2) is an interferon-inducible transmembrane protein that potently inhibits the release of nascent retrovirus particles. BST-2 is downregulated by the viral Vpu protein. BST-2 acts as a direct physical tether, which holds and links virions at the cell membrane. These tethered virons can remain on the cell surface or be internalized by endocytosis for degradation. BST-2 inhibits MMP2 and MMP15 activity, resulting in the attenuation of cellular proliferation and migration. BST-2 interaction with LILRA/ILT7 receptors in plasmacytoid dendritic cells negatively regulates IFN production in response to viral infection.
~33 kDa observed. The calculated molecular weight of this protein is 20 kDa, but can be observed at ~25-35 kDa due to high glycosylation.

Immunogen

Epitope: extracellular domain
KLH-conjugated linear peptide corresponding to the extracellular domain of human BST2.

Application

Anti-BST2 is an antibody targeting the BST2 protein, validated for use in WB & IHC.
Immunohistochemistry Analysis: A 1:500-2,000 dilution from a representative lot detected BST2 in human cardiac myocyte and human hepatocyte tissues.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Biochem/physiol Actions

This antibody recognizes the extracellular domain of BST2

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human bone marrow tissue lysate
Evaluated by Western Blot in human bone marrow tissue lysate.

Western Blot Analysis: 1.0 µg/mL of this antibody detected BST2 in 10 µg of human bone marrow tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: MABF201

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Venkata Subba Rao Atluri et al.
Scientific reports, 6, 27864-27864 (2016-06-21)
We have observed significantly increased HIV infection in HIV infected macrophages in the presence of cocaine that could be due to the downregulation of BST2 restriction factor in these cells. In human inflammasome PCR array, among different involved in inflammasome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service